Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 100258486-100258743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF19A
Entrez Gene ID
25897
Gene Name
ring finger protein 19A, RBR E3 ubiquitin protein ligase
Gene Aliases
RNF19
UniGene
Hs.292882
Gene Chromosome Location
Chr.8: 100257059-100336218 on Build GRCh38
Transcripts
NM_001280539.1 NM_015435.4 NM_183419.3 XM_005250853.3 XM_011516956.2 XM_017013302.1 XM_017013303.1 XM_017013304.1 XM_017013305.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3269107 c.2447T>C p.L816P Chr.8: 100258626-100258626 on Build GRCh38
748091 c.2389G>A p.E797K Chr.8: 100258684-100258684 on Build GRCh38
108826 c.2452G>A p.E818K Chr.8: 100258621-100258621 on Build GRCh38
5968463 c.2344C>A p.Q782K Chr.8: 100258729-100258729 on Build GRCh38
6767551 c.2342C>T p.T781I Chr.8: 100258731-100258731 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs931604114 Chr.8: 100258604-100258604 on Build GRCh38
rs752854364 Chr.8: 100258677-100258677 on Build GRCh38
rs376881086 Chr.8: 100258520-100258520 on Build GRCh38
rs985453783 Chr.8: 100258689-100258689 on Build GRCh38
rs567250945 Chr.8: 100258668-100258668 on Build GRCh38
rs1005707474 Chr.8: 100258519-100258519 on Build GRCh38
rs9642785 Chr.8: 100258568-100258568 on Build GRCh38
rs374144598 Chr.8: 100258713-100258713 on Build GRCh38
rs769692261 Chr.8: 100258593-100258593 on Build GRCh38
rs766890277 Chr.8: 100258639-100258639 on Build GRCh38
rs377389531 Chr.8: 100258595-100258595 on Build GRCh38
rs756342854 Chr.8: 100258696-100258696 on Build GRCh38
rs145880682 Chr.8: 100258621-100258621 on Build GRCh38
rs370795166 Chr.8: 100258614-100258614 on Build GRCh38
rs780781380 Chr.8: 100258547-100258547 on Build GRCh38
rs774326075 Chr.8: 100258611-100258611 on Build GRCh38
rs368066114 Chr.8: 100258702-100258702 on Build GRCh38
rs202182583 Chr.8: 100258552-100258552 on Build GRCh38
rs145481850 Chr.8: 100258517-100258517 on Build GRCh38
rs375014597 Chr.8: 100258633-100258633 on Build GRCh38
rs770482112 Chr.8: 100258543-100258543 on Build GRCh38
rs758935043 Chr.8: 100258539-100258539 on Build GRCh38
rs777781418 Chr.8: 100258700-100258700 on Build GRCh38
rs781339153 Chr.8: 100258590-100258590 on Build GRCh38
rs113974400 Chr.8: 100258530-100258530 on Build GRCh38
rs750838487 Chr.8: 100258537-100258537 on Build GRCh38
rs536162866 Chr.8: 100258703-100258703 on Build GRCh38
rs770449613 Chr.8: 100258512-100258512 on Build GRCh38
rs748256249 Chr.8: 100258592-100258592 on Build GRCh38
rs759327903 Chr.8: 100258518-100258518 on Build GRCh38
rs1003022684 Chr.8: 100258642-100258642 on Build GRCh38
rs35177403 Chr.8: 100258634-100258634 on Build GRCh38
rs527727281 Chr.8: 100258630-100258630 on Build GRCh38
rs775217409 Chr.8: 100258627-100258627 on Build GRCh38
rs771883804 Chr.8: 100258616-100258616 on Build GRCh38
rs765779046 Chr.8: 100258535-100258535 on Build GRCh38
rs77034032 Chr.8: 100258613-100258613 on Build GRCh38
rs751946324 Chr.8: 100258651-100258651 on Build GRCh38
rs755293164 Chr.8: 100258661-100258661 on Build GRCh38
rs749241840 Chr.8: 100258605-100258605 on Build GRCh38
rs773822737 Chr.8: 100258610-100258610 on Build GRCh38
rs751893153 Chr.8: 100258575-100258575 on Build GRCh38
rs745522197 Chr.8: 100258607-100258609 on Build GRCh38
rs760245590 Chr.8: 100258629-100258629 on Build GRCh38

View Full Product Details