Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 149943905-149944160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTUD7B
Entrez Gene ID
56957
Gene Name
OTU deubiquitinase 7B
Gene Aliases
CEZANNE, ZA20D1
UniGene
Hs.98322
Gene Chromosome Location
Chr.1: 149940330-150010766 on Build GRCh38
Transcripts
NM_020205.3 XM_011509782.1 XM_011509783.2 XM_011509784.2 XM_011509785.2 XM_011509786.1 XM_011509787.1 XM_011509788.1 XM_017001850.1 XM_017001851.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1196956 c.2378G>A p.R793Q Chr.1: 149944011-149944011 on Build GRCh38
6507708 c.2327G>T p.S776I Chr.1: 149944062-149944062 on Build GRCh38
6057943 c.2364G>T p.W788C Chr.1: 149944025-149944025 on Build GRCh38
5843520 c.2303C>T p.P768L Chr.1: 149944086-149944086 on Build GRCh38
5032034 c.2469C>A p.Y823* Chr.1: 149943920-149943920 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782580313 Chr.1: 149943986-149943986 on Build GRCh38
rs782317970 Chr.1: 149944087-149944087 on Build GRCh38
rs782451049 Chr.1: 149943980-149943980 on Build GRCh38
rs781835327 Chr.1: 149944136-149944136 on Build GRCh38
rs987090361 Chr.1: 149944139-149944139 on Build GRCh38
rs587752377 Chr.1: 149944081-149944081 on Build GRCh38
rs782025471 Chr.1: 149944043-149944043 on Build GRCh38
rs781882769 Chr.1: 149943945-149943945 on Build GRCh38
rs781802267 Chr.1: 149943932-149943932 on Build GRCh38
rs587649806 Chr.1: 149944101-149944101 on Build GRCh38
rs200128430 Chr.1: 149944012-149944012 on Build GRCh38
rs782429516 Chr.1: 149943935-149943935 on Build GRCh38
rs782616261 Chr.1: 149943974-149943974 on Build GRCh38
rs76350959 Chr.1: 149944134-149944134 on Build GRCh38
rs782407521 Chr.1: 149944001-149944001 on Build GRCh38
rs782574025 Chr.1: 149944088-149944088 on Build GRCh38
rs781887438 Chr.1: 149944128-149944128 on Build GRCh38
rs782213642 Chr.1: 149944086-149944086 on Build GRCh38
rs782283906 Chr.1: 149944089-149944089 on Build GRCh38
rs782470121 Chr.1: 149944137-149944137 on Build GRCh38
rs1028539832 Chr.1: 149944114-149944114 on Build GRCh38
rs782658838 Chr.1: 149944065-149944065 on Build GRCh38
rs782566215 Chr.1: 149944064-149944064 on Build GRCh38
rs782003109 Chr.1: 149944003-149944003 on Build GRCh38
rs377196376 Chr.1: 149943963-149943963 on Build GRCh38
rs782804183 Chr.1: 149943938-149943938 on Build GRCh38
rs782769953 Chr.1: 149944052-149944052 on Build GRCh38
rs782732204 Chr.1: 149944059-149944059 on Build GRCh38
rs782297708 Chr.1: 149943990-149943990 on Build GRCh38
rs781939731 Chr.1: 149944100-149944100 on Build GRCh38
rs954269727 Chr.1: 149944138-149944138 on Build GRCh38
rs782683888 Chr.1: 149944042-149944042 on Build GRCh38
rs782614848 Chr.1: 149944085-149944085 on Build GRCh38
rs367568103 Chr.1: 149944090-149944090 on Build GRCh38
rs267598003 Chr.1: 149943975-149943975 on Build GRCh38
rs782126031 Chr.1: 149944131-149944131 on Build GRCh38
rs1013492701 Chr.1: 149943962-149943962 on Build GRCh38
rs781793506 Chr.1: 149944060-149944060 on Build GRCh38
rs782512356 Chr.1: 149944040-149944041 on Build GRCh38
rs782641251 Chr.1: 149944075-149944075 on Build GRCh38
rs782061349 Chr.1: 149944037-149944037 on Build GRCh38
rs199673670 Chr.1: 149943960-149943960 on Build GRCh38
rs782473931 Chr.1: 149944058-149944058 on Build GRCh38
rs782804760 Chr.1: 149944111-149944111 on Build GRCh38
rs782263900 Chr.1: 149944073-149944073 on Build GRCh38
rs782141797 Chr.1: 149944046-149944046 on Build GRCh38
rs902208440 Chr.1: 149944007-149944007 on Build GRCh38
rs782222539 Chr.1: 149944123-149944124 on Build GRCh38

View Full Product Details