Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14451628-14451879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PKN1
Entrez Gene ID
5585
Gene Name
protein kinase N1
Gene Aliases
DBK, PAK-1, PAK1, PKN, PKN-ALPHA, PRK1, PRKCL1
UniGene
Hs.466044
Gene Chromosome Location
Chr.19: 14433354-14471867 on Build GRCh38
Transcripts
NM_002741.3 NM_213560.1 XM_011528128.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6747482 c.1014C>T p.G338G Chr.19: 14451854-14451854 on Build GRCh38
4457282 c.1039C>T p.P347S Chr.19: 14451879-14451879 on Build GRCh38
72195 c.980A>G p.E327G Chr.19: 14451838-14451838 on Build GRCh38
5620062 c.1021G>T p.D341Y Chr.19: 14451861-14451861 on Build GRCh38
3529343 c.1009C>T p.P337S Chr.19: 14451867-14451867 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779081221 Chr.19: 14451837-14451837 on Build GRCh38
rs946668645 Chr.19: 14451691-14451691 on Build GRCh38
rs567156865 Chr.19: 14451736-14451736 on Build GRCh38
rs780176880 Chr.19: 14451817-14451817 on Build GRCh38
rs542773853 Chr.19: 14451833-14451833 on Build GRCh38
rs371450588 Chr.19: 14451658-14451658 on Build GRCh38
rs8111147 Chr.19: 14451774-14451774 on Build GRCh38
rs887206450 Chr.19: 14451735-14451735 on Build GRCh38
rs553819615 Chr.19: 14451809-14451809 on Build GRCh38
rs180998014 Chr.19: 14451702-14451702 on Build GRCh38
rs1004665055 Chr.19: 14451741-14451741 on Build GRCh38
rs73924184 Chr.19: 14451713-14451713 on Build GRCh38
rs368109481 Chr.19: 14451764-14451764 on Build GRCh38
rs376876364 Chr.19: 14451836-14451836 on Build GRCh38
rs1031806748 Chr.19: 14451791-14451791 on Build GRCh38
rs1012904436 Chr.19: 14451773-14451773 on Build GRCh38
rs767244624 Chr.19: 14451804-14451804 on Build GRCh38
rs756865147 Chr.19: 14451802-14451802 on Build GRCh38
rs544934065 Chr.19: 14451797-14451797 on Build GRCh38
rs749236976 Chr.19: 14451823-14451823 on Build GRCh38
rs188188590 Chr.19: 14451679-14451679 on Build GRCh38
rs909196183 Chr.19: 14451760-14451760 on Build GRCh38
rs766861943 Chr.19: 14451779-14451779 on Build GRCh38
rs937562725 Chr.19: 14451731-14451731 on Build GRCh38
rs143159370 Chr.19: 14451778-14451778 on Build GRCh38
rs748326319 Chr.19: 14451842-14451842 on Build GRCh38
rs560950919 Chr.19: 14451850-14451850 on Build GRCh38
rs551758525 Chr.19: 14451686-14451686 on Build GRCh38
rs772298530 Chr.19: 14451843-14451843 on Build GRCh38
rs957055575 Chr.19: 14451670-14451670 on Build GRCh38
rs898459523 Chr.19: 14451763-14451763 on Build GRCh38
rs200776092 Chr.19: 14451789-14451789 on Build GRCh38
rs372582175 Chr.19: 14451799-14451799 on Build GRCh38
rs74755157 Chr.19: 14451693-14451693 on Build GRCh38
rs375528110 Chr.19: 14451661-14451661 on Build GRCh38
rs1050875772 Chr.19: 14451732-14451732 on Build GRCh38
rs971178052 Chr.19: 14451697-14451697 on Build GRCh38
rs186242949 Chr.19: 14451810-14451810 on Build GRCh38
rs1016952476 Chr.19: 14451742-14451742 on Build GRCh38
rs747450809 Chr.19: 14451854-14451854 on Build GRCh38

View Full Product Details