Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6743351-6743608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIP10
Entrez Gene ID
9322
Gene Name
thyroid hormone receptor interactor 10
Gene Aliases
CIP4, HSTP, STOT, STP, TRIP-10
UniGene
Hs.515094
Gene Chromosome Location
Chr.19: 6739682-6751526 on Build GRCh38
Transcripts
NM_001288962.1 NM_001288963.1 NM_004240.3 NR_110231.1 XM_005259683.2 XM_006722940.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
316180 c.486C>G p.I162M Chr.19: 6743571-6743571 on Build GRCh38
4082631 c.490G>A p.A164T Chr.19: 6743575-6743575 on Build GRCh38
4082630 c.490G>A p.A164T Chr.19: 6743575-6743575 on Build GRCh38
4082628 c.434G>A p.C145Y Chr.19: 6743519-6743519 on Build GRCh38
4828427 c.424G>C p.E142Q Chr.19: 6743509-6743509 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984059691 Chr.19: 6743441-6743441 on Build GRCh38
rs381897 Chr.19: 6743388-6743388 on Build GRCh38
rs561466455 Chr.19: 6743436-6743436 on Build GRCh38
rs373417130 Chr.19: 6743501-6743501 on Build GRCh38
rs766852682 Chr.19: 6743483-6743483 on Build GRCh38
rs770309049 Chr.19: 6743573-6743573 on Build GRCh38
rs759979487 Chr.19: 6743486-6743486 on Build GRCh38
rs140217591 Chr.19: 6743380-6743380 on Build GRCh38
rs986262259 Chr.19: 6743456-6743456 on Build GRCh38
rs145688036 Chr.19: 6743554-6743554 on Build GRCh38
rs376741923 Chr.19: 6743521-6743521 on Build GRCh38
rs774942232 Chr.19: 6743574-6743574 on Build GRCh38
rs918186324 Chr.19: 6743433-6743433 on Build GRCh38
rs556421398 Chr.19: 6743446-6743446 on Build GRCh38
rs767925174 Chr.19: 6743579-6743579 on Build GRCh38
rs540346498 Chr.19: 6743555-6743555 on Build GRCh38
rs570469259 Chr.19: 6743420-6743420 on Build GRCh38
rs145227710 Chr.19: 6743475-6743475 on Build GRCh38
rs780202082 Chr.19: 6743522-6743523 on Build GRCh38
rs759899109 Chr.19: 6743575-6743575 on Build GRCh38
rs149097976 Chr.19: 6743522-6743522 on Build GRCh38
rs761315005 Chr.19: 6743473-6743473 on Build GRCh38
rs144976733 Chr.19: 6743500-6743500 on Build GRCh38
rs763513711 Chr.19: 6743479-6743479 on Build GRCh38
rs751912954 Chr.19: 6743485-6743485 on Build GRCh38
rs769298439 Chr.19: 6743474-6743474 on Build GRCh38
rs758457903 Chr.19: 6743512-6743512 on Build GRCh38
rs762422168 Chr.19: 6743566-6743566 on Build GRCh38
rs752974350 Chr.19: 6743497-6743497 on Build GRCh38
rs1038774617 Chr.19: 6743480-6743480 on Build GRCh38
rs747863740 Chr.19: 6743526-6743526 on Build GRCh38
rs200899289 Chr.19: 6743454-6743454 on Build GRCh38
rs746796230 Chr.19: 6743449-6743449 on Build GRCh38
rs2277986 Chr.19: 6743394-6743394 on Build GRCh38
rs746874869 Chr.19: 6743515-6743515 on Build GRCh38
rs780009125 Chr.19: 6743513-6743513 on Build GRCh38
rs891743647 Chr.19: 6743545-6743545 on Build GRCh38
rs1030493290 Chr.19: 6743428-6743428 on Build GRCh38
rs776370872 Chr.19: 6743470-6743470 on Build GRCh38
rs767722879 Chr.19: 6743488-6743488 on Build GRCh38

View Full Product Details