Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 146459901-146460130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLSCR2
Entrez Gene ID
57047
Gene Name
phospholipid scramblase 2
Gene Aliases
-
UniGene
Hs.744414
Gene Chromosome Location
Chr.3: 146391363-146497163 on Build GRCh38
Transcripts
NM_001199978.1 NM_001199979.1 NM_020359.2 XM_011513013.2 XM_011513019.2 XM_011513020.2 XM_011513021.2 XM_011513022.2 XM_011513023.2 XM_017006898.1 XM_017006899.1 XM_017006900.1 XM_017006901.1 XM_017006902.1 XM_017006903.1 XM_017006904.1 XM_017006905.1 XM_017006906.1 XM_017006907.1 XM_017006908.1 XM_017006909.1 XM_017006910.1 XM_017006911.1 XM_017006912.1 XM_017006913.1 XM_017006914.1 XM_017006915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3846454 c.96G>A p.Q32Q Chr.3: 146460028-146460028 on Build GRCh38
1221227 c.3G>A p.M1I Chr.3: 146459902-146459902 on Build GRCh38
1693562 c.178C>T p.P60S Chr.3: 146459946-146459946 on Build GRCh38
6219086 c.133C>T p.R45C Chr.3: 146459991-146459991 on Build GRCh38
3944967 c.50G>A p.G17E Chr.3: 146460074-146460074 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs75562053 Chr.3: 146460064-146460064 on Build GRCh38
rs959891544 Chr.3: 146460062-146460062 on Build GRCh38
rs574913560 Chr.3: 146459991-146459991 on Build GRCh38
rs765210100 Chr.3: 146459957-146459957 on Build GRCh38
rs768297391 Chr.3: 146460069-146460069 on Build GRCh38
rs6806535 Chr.3: 146460028-146460028 on Build GRCh38
rs772729755 Chr.3: 146460100-146460100 on Build GRCh38
rs770918033 Chr.3: 146460076-146460076 on Build GRCh38
rs571600120 Chr.3: 146459990-146459990 on Build GRCh38
rs778314395 Chr.3: 146460021-146460021 on Build GRCh38
rs776140560 Chr.3: 146459946-146459946 on Build GRCh38
rs768228340 Chr.3: 146459940-146459940 on Build GRCh38
rs762713442 Chr.3: 146459981-146459981 on Build GRCh38
rs374893715 Chr.3: 146460075-146460075 on Build GRCh38
rs564375368 Chr.3: 146460101-146460101 on Build GRCh38
rs1031492856 Chr.3: 146460053-146460053 on Build GRCh38
rs773060953 Chr.3: 146459980-146459980 on Build GRCh38
rs756710377 Chr.3: 146459938-146459938 on Build GRCh38
rs933241316 Chr.3: 146460066-146460066 on Build GRCh38
rs776206847 Chr.3: 146460071-146460071 on Build GRCh38
rs752804862 Chr.3: 146460010-146460010 on Build GRCh38
rs756212569 Chr.3: 146460015-146460015 on Build GRCh38
rs747691324 Chr.3: 146460076-146460076 on Build GRCh38
rs769235764 Chr.3: 146460085-146460085 on Build GRCh38
rs535945285 Chr.3: 146460003-146460003 on Build GRCh38
rs966806265 Chr.3: 146459973-146459973 on Build GRCh38
rs572425835 Chr.3: 146460059-146460059 on Build GRCh38
rs749862029 Chr.3: 146460022-146460022 on Build GRCh38
rs998667622 Chr.3: 146460051-146460051 on Build GRCh38
rs149831493 Chr.3: 146459949-146459949 on Build GRCh38
rs779385687 Chr.3: 146460034-146460034 on Build GRCh38

View Full Product Details