Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 104772913-104773138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NIPSNAP3B
Entrez Gene ID
55335
Gene Name
nipsnap homolog 3B
Gene Aliases
FP944, NIPSNAP3, SNAP1
UniGene
Hs.429294
Gene Chromosome Location
Chr.9: 104763741-104777764 on Build GRCh38
Transcripts
NM_018376.3 NR_130759.1 NR_130760.1 XM_011518839.1 XM_017014889.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5625384 c.702G>A p.Q234Q Chr.9: 104773031-104773031 on Build GRCh38
5567761 c.695C>A p.S232Y Chr.9: 104773024-104773024 on Build GRCh38
3902732 c.726A>G p.S242S Chr.9: 104773055-104773055 on Build GRCh38
1103511 c.693T>A p.V231V Chr.9: 104773022-104773022 on Build GRCh38
1103510 c.670C>T p.R224W Chr.9: 104772999-104772999 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778348014 Chr.9: 104772984-104772984 on Build GRCh38
rs377506623 Chr.9: 104772962-104772962 on Build GRCh38
rs759656734 Chr.9: 104773015-104773015 on Build GRCh38
rs768505634 Chr.9: 104773060-104773060 on Build GRCh38
rs764481365 Chr.9: 104773023-104773023 on Build GRCh38
rs529659668 Chr.9: 104773003-104773003 on Build GRCh38
rs779445447 Chr.9: 104773052-104773052 on Build GRCh38
rs769748783 Chr.9: 104773072-104773072 on Build GRCh38
rs552388877 Chr.9: 104773089-104773089 on Build GRCh38
rs957808488 Chr.9: 104772995-104772995 on Build GRCh38
rs754178768 Chr.9: 104773027-104773027 on Build GRCh38
rs772219262 Chr.9: 104773100-104773100 on Build GRCh38
rs775435890 Chr.9: 104773111-104773111 on Build GRCh38
rs767772776 Chr.9: 104773018-104773018 on Build GRCh38
rs779967255 Chr.9: 104773000-104773000 on Build GRCh38
rs780989418 Chr.9: 104773063-104773063 on Build GRCh38
rs753232095 Chr.9: 104773020-104773020 on Build GRCh38
rs750147301 Chr.9: 104773038-104773038 on Build GRCh38
rs150554330 Chr.9: 104772999-104772999 on Build GRCh38
rs750851064 Chr.9: 104773029-104773029 on Build GRCh38
rs756325897 Chr.9: 104772982-104772982 on Build GRCh38
rs562267988 Chr.9: 104772956-104772956 on Build GRCh38
rs149753675 Chr.9: 104773022-104773022 on Build GRCh38
rs778056091 Chr.9: 104772964-104772964 on Build GRCh38
rs771297073 Chr.9: 104773007-104773007 on Build GRCh38
rs745360995 Chr.9: 104772996-104772996 on Build GRCh38
rs750456883 Chr.9: 104772978-104772978 on Build GRCh38
rs774759408 Chr.9: 104773009-104773009 on Build GRCh38
rs753471162 Chr.9: 104772976-104772976 on Build GRCh38
rs780408589 Chr.9: 104772986-104772986 on Build GRCh38
rs756892647 Chr.9: 104772979-104772979 on Build GRCh38
rs368271075 Chr.9: 104773078-104773078 on Build GRCh38
rs746900711 Chr.9: 104773059-104773059 on Build GRCh38
rs778507463 Chr.9: 104773034-104773034 on Build GRCh38
rs747869885 Chr.9: 104773064-104773064 on Build GRCh38
rs763192312 Chr.9: 104773006-104773006 on Build GRCh38
rs140617020 Chr.9: 104773048-104773048 on Build GRCh38
rs56270385 Chr.9: 104773004-104773004 on Build GRCh38

View Full Product Details