Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119357670-119357923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP2
Entrez Gene ID
9099
Gene Name
ubiquitin specific peptidase 2
Gene Aliases
UBP41, USP9
UniGene
Hs.524085
Gene Chromosome Location
Chr.11: 119355215-119381726 on Build GRCh38
Transcripts
NM_001243759.1 NM_004205.4 NM_171997.2 XM_005271721.4 XM_005271722.2 XM_017018539.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7222291 c.796-1G>A p.? Chr.11: 119357836-119357836 on Build GRCh38
5999876 c.828T>C p.C276C Chr.11: 119357803-119357803 on Build GRCh38
1585449 c.836A>C p.K279T Chr.11: 119357795-119357795 on Build GRCh38
4610142 c.1432C>T p.R478C Chr.11: 119357826-119357826 on Build GRCh38
3676002 c.1480T>G p.F494V Chr.11: 119357778-119357778 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746737928 Chr.11: 119357887-119357887 on Build GRCh38
rs765470322 Chr.11: 119357848-119357851 on Build GRCh38
rs397897488 Chr.11: 119357842-119357843 on Build GRCh38
rs763624175 Chr.11: 119357826-119357826 on Build GRCh38
rs752940333 Chr.11: 119357859-119357859 on Build GRCh38
rs780042046 Chr.11: 119357885-119357885 on Build GRCh38
rs745844862 Chr.11: 119357778-119357778 on Build GRCh38
rs762800168 Chr.11: 119357882-119357882 on Build GRCh38
rs45619639 Chr.11: 119357838-119357839 on Build GRCh38
rs781491629 Chr.11: 119357758-119357758 on Build GRCh38
rs769475367 Chr.11: 119357817-119357817 on Build GRCh38
rs751816503 Chr.11: 119357830-119357830 on Build GRCh38
rs759950930 Chr.11: 119357838-119357838 on Build GRCh38
rs748109847 Chr.11: 119357760-119357760 on Build GRCh38
rs771847802 Chr.11: 119357790-119357790 on Build GRCh38
rs766697552 Chr.11: 119357746-119357746 on Build GRCh38
rs756166063 Chr.11: 119357864-119357864 on Build GRCh38
rs761356157 Chr.11: 119357808-119357808 on Build GRCh38
rs1033591051 Chr.11: 119357692-119357692 on Build GRCh38
rs199767133 Chr.11: 119357855-119357855 on Build GRCh38
rs746833172 Chr.11: 119357804-119357804 on Build GRCh38
rs750345664 Chr.11: 119357880-119357880 on Build GRCh38
rs767594325 Chr.11: 119357842-119357842 on Build GRCh38
rs1010409303 Chr.11: 119357737-119357737 on Build GRCh38
rs758210711 Chr.11: 119357881-119357881 on Build GRCh38
rs972996803 Chr.11: 119357693-119357693 on Build GRCh38
rs1029795266 Chr.11: 119357699-119357699 on Build GRCh38
rs566621045 Chr.11: 119357807-119357807 on Build GRCh38
rs61753081 Chr.11: 119357819-119357819 on Build GRCh38
rs971346165 Chr.11: 119357810-119357810 on Build GRCh38
rs775206449 Chr.11: 119357800-119357800 on Build GRCh38
rs980605669 Chr.11: 119357818-119357818 on Build GRCh38
rs768123790 Chr.11: 119357806-119357806 on Build GRCh38
rs371245887 Chr.11: 119357749-119357749 on Build GRCh38
rs771520695 Chr.11: 119357723-119357723 on Build GRCh38
rs756219325 Chr.11: 119357769-119357769 on Build GRCh38
rs200595852 Chr.11: 119357843-119357844 on Build GRCh38
rs189608025 Chr.11: 119357827-119357827 on Build GRCh38
rs377758729 Chr.11: 119357714-119357714 on Build GRCh38
rs778775200 Chr.11: 119357770-119357770 on Build GRCh38
rs755216327 Chr.11: 119357756-119357756 on Build GRCh38
rs758816784 Chr.11: 119357727-119357727 on Build GRCh38
rs952226623 Chr.11: 119357847-119357847 on Build GRCh38
rs911293045 Chr.11: 119357802-119357802 on Build GRCh38
rs374949575 Chr.11: 119357879-119357879 on Build GRCh38

View Full Product Details