Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 103079786-103080043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NXF3
Entrez Gene ID
56000
Gene Name
nuclear RNA export factor 3
Gene Aliases
-
UniGene
Hs.60386
Gene Chromosome Location
Chr.X: 103075821-103093094 on Build GRCh38
Transcripts
NM_022052.1

SNP Detail

SNP ID Chromosome Location
rs768620043 Chr.X: 103079974-103079974 on Build GRCh38
rs750599908 Chr.X: 103079816-103079816 on Build GRCh38
rs185213131 Chr.X: 103079905-103079905 on Build GRCh38
rs921381780 Chr.X: 103079909-103079909 on Build GRCh38
rs774332099 Chr.X: 103079993-103079993 on Build GRCh38
rs369030730 Chr.X: 103079815-103079815 on Build GRCh38
rs775025954 Chr.X: 103079921-103079921 on Build GRCh38
rs756761095 Chr.X: 103079897-103079897 on Build GRCh38
rs375878656 Chr.X: 103079889-103079889 on Build GRCh38
rs761699474 Chr.X: 103080010-103080010 on Build GRCh38
rs755460265 Chr.X: 103079842-103079842 on Build GRCh38
rs767912387 Chr.X: 103080017-103080017 on Build GRCh38
rs780112424 Chr.X: 103079837-103079837 on Build GRCh38
rs745465990 Chr.X: 103079904-103079904 on Build GRCh38
rs868104968 Chr.X: 103079931-103079931 on Build GRCh38
rs756273075 Chr.X: 103079835-103079835 on Build GRCh38
rs749205500 Chr.X: 103079969-103079969 on Build GRCh38
rs372588184 Chr.X: 103079841-103079841 on Build GRCh38
rs144925686 Chr.X: 103079862-103079862 on Build GRCh38
rs770635481 Chr.X: 103079873-103079873 on Build GRCh38
rs1016029463 Chr.X: 103079872-103079872 on Build GRCh38
rs762178262 Chr.X: 103079807-103079807 on Build GRCh38
rs780635414 Chr.X: 103079898-103079898 on Build GRCh38
rs779195104 Chr.X: 103079854-103079854 on Build GRCh38
rs748481077 Chr.X: 103079858-103079858 on Build GRCh38
rs773569670 Chr.X: 103080018-103080018 on Build GRCh38

View Full Product Details