Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24502027-24502300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UPB1
Entrez Gene ID
51733
Gene Name
beta-ureidopropionase 1
Gene Aliases
BUP1
UniGene
Hs.731656
Gene Chromosome Location
Chr.22: 24495060-24528681 on Build GRCh38
Transcripts
NM_016327.2 XM_011530222.2 XM_011530223.2 XM_011530224.2 XM_011530225.2 XM_017028825.1 XM_017028826.1 XM_017028827.1 XM_017028828.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6273477 c.319G>T p.A107S Chr.22: 24502168-24502168 on Build GRCh38
7258873 c.358G>A p.A120T Chr.22: 24502207-24502207 on Build GRCh38
1032716 c.290A>G p.H97R Chr.22: 24502139-24502139 on Build GRCh38
419620 c.296G>A p.R99H Chr.22: 24502145-24502145 on Build GRCh38
4640398 c.282T>C p.S94S Chr.22: 24502131-24502131 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762325580 Chr.22: 24502223-24502223 on Build GRCh38
rs61250014 Chr.22: 24502095-24502095 on Build GRCh38
rs773263627 Chr.22: 24502253-24502253 on Build GRCh38
rs903878408 Chr.22: 24502102-24502102 on Build GRCh38
rs751120275 Chr.22: 24502135-24502135 on Build GRCh38
rs750057554 Chr.22: 24502145-24502145 on Build GRCh38
rs780958577 Chr.22: 24502144-24502144 on Build GRCh38
rs770926451 Chr.22: 24502168-24502168 on Build GRCh38
rs774843626 Chr.22: 24502139-24502139 on Build GRCh38
rs749228384 Chr.22: 24502075-24502075 on Build GRCh38
rs754750827 Chr.22: 24502256-24502256 on Build GRCh38
rs770091394 Chr.22: 24502203-24502203 on Build GRCh38
rs776702958 Chr.22: 24502170-24502170 on Build GRCh38
rs778793828 Chr.22: 24502158-24502158 on Build GRCh38
rs956484720 Chr.22: 24502206-24502206 on Build GRCh38
rs976761982 Chr.22: 24502252-24502252 on Build GRCh38
rs767223788 Chr.22: 24502139-24502140 on Build GRCh38
rs1045062028 Chr.22: 24502052-24502052 on Build GRCh38
rs773305083 Chr.22: 24502079-24502079 on Build GRCh38
rs746105287 Chr.22: 24502175-24502175 on Build GRCh38
rs753071301 Chr.22: 24502122-24502122 on Build GRCh38
rs755693951 Chr.22: 24502205-24502205 on Build GRCh38
rs760995440 Chr.22: 24502271-24502271 on Build GRCh38
rs528445172 Chr.22: 24502049-24502049 on Build GRCh38
rs202094164 Chr.22: 24502124-24502124 on Build GRCh38
rs772212502 Chr.22: 24502219-24502219 on Build GRCh38
rs763376783 Chr.22: 24502213-24502213 on Build GRCh38
rs765696813 Chr.22: 24502109-24502109 on Build GRCh38
rs749883671 Chr.22: 24502188-24502190 on Build GRCh38
rs1019345080 Chr.22: 24502214-24502214 on Build GRCh38
rs760651191 Chr.22: 24502087-24502087 on Build GRCh38
rs778709015 Chr.22: 24502265-24502265 on Build GRCh38
rs755552957 Chr.22: 24502264-24502265 on Build GRCh38
rs138872001 Chr.22: 24502207-24502207 on Build GRCh38
rs749037327 Chr.22: 24502156-24502156 on Build GRCh38
rs1000662421 Chr.22: 24502194-24502194 on Build GRCh38
rs776701916 Chr.22: 24502089-24502089 on Build GRCh38
rs764763105 Chr.22: 24502218-24502218 on Build GRCh38
rs368072734 Chr.22: 24502229-24502229 on Build GRCh38
rs149380394 Chr.22: 24502088-24502088 on Build GRCh38
rs765997355 Chr.22: 24502239-24502239 on Build GRCh38
rs755789447 Chr.22: 24502233-24502233 on Build GRCh38
rs764675102 Chr.22: 24502128-24502128 on Build GRCh38
rs140157113 Chr.22: 24502201-24502201 on Build GRCh38
rs768631977 Chr.22: 24502157-24502157 on Build GRCh38
rs779859166 Chr.22: 24502152-24502152 on Build GRCh38
rs780416829 Chr.22: 24502221-24502221 on Build GRCh38
rs755697579 Chr.22: 24502147-24502147 on Build GRCh38
rs768545897 Chr.22: 24502078-24502078 on Build GRCh38
rs756719003 Chr.22: 24502140-24502140 on Build GRCh38
rs766941831 Chr.22: 24502227-24502227 on Build GRCh38
rs975590315 Chr.22: 24502278-24502278 on Build GRCh38
rs754450964 Chr.22: 24502220-24502221 on Build GRCh38

View Full Product Details