Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 69281003-69281260 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UGT2B28
Entrez Gene ID
54490
Gene Name
UDP glucuronosyltransferase family 2 member B28
Gene Aliases
-
UniGene
Hs.653154
Gene Chromosome Location
Chr.4: 69280429-69295050 on Build GRCh38
Transcripts
NM_001207004.1 NM_053039.1 XM_017008303.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3605261 c.541G>A p.E181K Chr.4: 69281041-69281041 on Build GRCh38
3605263 c.687G>A p.M229I Chr.4: 69281187-69281187 on Build GRCh38
2829602 c.545G>A p.R182K Chr.4: 69281045-69281045 on Build GRCh38
3605262 c.584C>T p.P195L Chr.4: 69281084-69281084 on Build GRCh38
3605265 c.712G>A p.E238K Chr.4: 69281212-69281212 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764085145 Chr.4: 69281045-69281045 on Build GRCh38
rs767756470 Chr.4: 69281201-69281201 on Build GRCh38
rs868351917 Chr.4: 69281178-69281192 on Build GRCh38
rs148987832 Chr.4: 69281086-69281086 on Build GRCh38
rs768747813 Chr.4: 69281170-69281170 on Build GRCh38
rs747723755 Chr.4: 69281167-69281167 on Build GRCh38
rs201064783 Chr.4: 69281083-69281083 on Build GRCh38
rs746719102 Chr.4: 69281177-69281177 on Build GRCh38
rs779238247 Chr.4: 69281164-69281164 on Build GRCh38
rs761045559 Chr.4: 69281213-69281213 on Build GRCh38
rs138591037 Chr.4: 69281184-69281184 on Build GRCh38
rs575693723 Chr.4: 69281152-69281152 on Build GRCh38
rs770562090 Chr.4: 69281178-69281178 on Build GRCh38
rs757491604 Chr.4: 69281062-69281063 on Build GRCh38
rs772813538 Chr.4: 69281105-69281105 on Build GRCh38
rs879922056 Chr.4: 69281062-69281062 on Build GRCh38
rs780534422 Chr.4: 69281084-69281084 on Build GRCh38
rs781210281 Chr.4: 69281068-69281068 on Build GRCh38
rs369845280 Chr.4: 69281211-69281211 on Build GRCh38
rs573870440 Chr.4: 69281134-69281134 on Build GRCh38
rs766214321 Chr.4: 69281113-69281113 on Build GRCh38
rs762918907 Chr.4: 69281186-69281186 on Build GRCh38
rs757501499 Chr.4: 69281053-69281053 on Build GRCh38
rs200358445 Chr.4: 69281092-69281092 on Build GRCh38
rs1030550795 Chr.4: 69281200-69281200 on Build GRCh38
rs373101039 Chr.4: 69281123-69281123 on Build GRCh38
rs767885815 Chr.4: 69281132-69281132 on Build GRCh38
rs760292004 Chr.4: 69281108-69281108 on Build GRCh38
rs375204354 Chr.4: 69281116-69281116 on Build GRCh38
rs754251721 Chr.4: 69281215-69281215 on Build GRCh38
rs201982815 Chr.4: 69281172-69281172 on Build GRCh38
rs751583259 Chr.4: 69281049-69281049 on Build GRCh38
rs756383423 Chr.4: 69281143-69281143 on Build GRCh38
rs759197918 Chr.4: 69281182-69281182 on Build GRCh38
rs765401221 Chr.4: 69281207-69281207 on Build GRCh38
rs775253593 Chr.4: 69281185-69281185 on Build GRCh38
rs757238220 Chr.4: 69281130-69281130 on Build GRCh38
rs558174566 Chr.4: 69281036-69281036 on Build GRCh38
rs773916411 Chr.4: 69281088-69281088 on Build GRCh38
rs369671261 Chr.4: 69281118-69281118 on Build GRCh38
rs766649977 Chr.4: 69281214-69281214 on Build GRCh38
rs746254363 Chr.4: 69281071-69281071 on Build GRCh38
rs375069193 Chr.4: 69281145-69281145 on Build GRCh38
rs554070311 Chr.4: 69281120-69281120 on Build GRCh38
rs753855746 Chr.4: 69281063-69281063 on Build GRCh38
rs755522570 Chr.4: 69281157-69281157 on Build GRCh38
rs1029518387 Chr.4: 69281121-69281121 on Build GRCh38
rs542781153 Chr.4: 69281139-69281139 on Build GRCh38
rs144771515 Chr.4: 69281180-69281180 on Build GRCh38
rs771779407 Chr.4: 69281104-69281104 on Build GRCh38
rs866054381 Chr.4: 69281061-69281061 on Build GRCh38

View Full Product Details