Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47582052-47582325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDUFS3
Entrez Gene ID
4722
Gene Name
NADH:ubiquinone oxidoreductase core subunit S3
Gene Aliases
CI-30
UniGene
Hs.502528
Gene Chromosome Location
Chr.11: 47579010-47584563 on Build GRCh38
Transcripts
NM_004551.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6068669 c.421A>G p.I141V Chr.11: 47582127-47582127 on Build GRCh38
5477592 c.406C>T p.R136C Chr.11: 47582112-47582112 on Build GRCh38
290066 c.456C>G p.P152P Chr.11: 47582162-47582162 on Build GRCh38
4424822 c.456C>T p.P152P Chr.11: 47582162-47582162 on Build GRCh38
340342 c.411C>T p.F137F Chr.11: 47582117-47582117 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759164951 Chr.11: 47582171-47582171 on Build GRCh38
rs774019045 Chr.11: 47582167-47582167 on Build GRCh38
rs368446373 Chr.11: 47582112-47582112 on Build GRCh38
rs770264683 Chr.11: 47582245-47582245 on Build GRCh38
rs775191064 Chr.11: 47582233-47582246 on Build GRCh38
rs749615057 Chr.11: 47582251-47582251 on Build GRCh38
rs748663281 Chr.11: 47582240-47582240 on Build GRCh38
rs142248674 Chr.11: 47582124-47582124 on Build GRCh38
rs540213433 Chr.11: 47582164-47582164 on Build GRCh38
rs770110842 Chr.11: 47582152-47582152 on Build GRCh38
rs771175881 Chr.11: 47582254-47582254 on Build GRCh38
rs754995990 Chr.11: 47582107-47582107 on Build GRCh38
rs374665190 Chr.11: 47582298-47582298 on Build GRCh38
rs778029739 Chr.11: 47582246-47582246 on Build GRCh38
rs372417584 Chr.11: 47582125-47582125 on Build GRCh38
rs756003708 Chr.11: 47582274-47582274 on Build GRCh38
rs368557574 Chr.11: 47582223-47582223 on Build GRCh38
rs751046043 Chr.11: 47582089-47582089 on Build GRCh38
rs753606950 Chr.11: 47582211-47582211 on Build GRCh38
rs148331180 Chr.11: 47582181-47582181 on Build GRCh38
rs767511454 Chr.11: 47582108-47582108 on Build GRCh38
rs759260043 Chr.11: 47582262-47582262 on Build GRCh38
rs777160160 Chr.11: 47582141-47582141 on Build GRCh38
rs375077765 Chr.11: 47582301-47582303 on Build GRCh38
rs144217602 Chr.11: 47582202-47582202 on Build GRCh38
rs763809636 Chr.11: 47582205-47582205 on Build GRCh38
rs755115327 Chr.11: 47582228-47582228 on Build GRCh38
rs564717508 Chr.11: 47582285-47582285 on Build GRCh38
rs762650545 Chr.11: 47582074-47582074 on Build GRCh38
rs769336340 Chr.11: 47582076-47582077 on Build GRCh38
rs748646798 Chr.11: 47582151-47582151 on Build GRCh38
rs146407178 Chr.11: 47582130-47582130 on Build GRCh38
rs774184651 Chr.11: 47582260-47582260 on Build GRCh38
rs748928602 Chr.11: 47582300-47582301 on Build GRCh38
rs113851567 Chr.11: 47582159-47582159 on Build GRCh38
rs2233358 Chr.11: 47582275-47582275 on Build GRCh38
rs28939714 Chr.11: 47582140-47582140 on Build GRCh38
rs75447314 Chr.11: 47582300-47582300 on Build GRCh38
rs1050853367 Chr.11: 47582276-47582276 on Build GRCh38
rs200601876 Chr.11: 47582293-47582293 on Build GRCh38
rs781231901 Chr.11: 47582235-47582235 on Build GRCh38
rs11552583 Chr.11: 47582192-47582192 on Build GRCh38
rs780005953 Chr.11: 47582131-47582131 on Build GRCh38
rs199783134 Chr.11: 47582184-47582184 on Build GRCh38
rs773564480 Chr.11: 47582158-47582158 on Build GRCh38
rs902177734 Chr.11: 47582268-47582268 on Build GRCh38
rs768879782 Chr.11: 47582229-47582229 on Build GRCh38
rs767197716 Chr.11: 47582189-47582189 on Build GRCh38
rs747054142 Chr.11: 47582138-47582138 on Build GRCh38
rs765816274 Chr.11: 47582076-47582076 on Build GRCh38
rs752656776 Chr.11: 47582111-47582111 on Build GRCh38
rs1055717968 Chr.11: 47582198-47582198 on Build GRCh38
rs371375887 Chr.11: 47582224-47582224 on Build GRCh38

View Full Product Details