Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35107271-35107532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD51D
Entrez Gene ID
5892
Gene Name
RAD51 paralog D
Gene Aliases
BROVCA4, R51H3, RAD51L3, TRAD
UniGene
Hs.631757
Gene Chromosome Location
Chr.17: 35099792-35119869 on Build GRCh38
Transcripts
NM_001142571.1 NM_002878.3 NM_133629.2 NR_037711.1 NR_037712.1 NR_037714.1

Gene Symbol
RAD51L3-RFFL
Entrez Gene ID
100529207
Gene Name
RAD51L3-RFFL readthrough
Gene Aliases
-
UniGene
Hs.13680
Gene Chromosome Location
Chr.17: 35009112-35121522 on Build GRCh38
Transcripts
NM_001142571.1 NM_002878.3 NM_133629.2 NR_037711.1 NR_037712.1 NR_037714.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5657737 c.380G>T p.G127V Chr.17: 35107391-35107391 on Build GRCh38
314660 c.320G>T p.G107V Chr.17: 35107391-35107391 on Build GRCh38
1630145 c.345+3_345+6delACAT p.? Chr.17: 35107360-35107363 on Build GRCh38
5055381 c.339T>C p.L113L Chr.17: 35107432-35107432 on Build GRCh38
7097226 c.371A>G p.E124G Chr.17: 35107400-35107400 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751293323 Chr.17: 35107496-35107496 on Build GRCh38
rs757481281 Chr.17: 35107461-35107461 on Build GRCh38
rs758132417 Chr.17: 35107387-35107387 on Build GRCh38
rs113147933 Chr.17: 35107426-35107426 on Build GRCh38
rs746416079 Chr.17: 35107416-35107416 on Build GRCh38
rs918947511 Chr.17: 35107362-35107362 on Build GRCh38
rs377572503 Chr.17: 35107486-35107486 on Build GRCh38
rs34678099 Chr.17: 35107297-35107298 on Build GRCh38
rs753529790 Chr.17: 35107454-35107454 on Build GRCh38
rs28363272 Chr.17: 35107481-35107481 on Build GRCh38
rs878854562 Chr.17: 35107361-35107361 on Build GRCh38
rs746648963 Chr.17: 35107398-35107398 on Build GRCh38
rs775651936 Chr.17: 35107424-35107424 on Build GRCh38
rs730882119 Chr.17: 35107384-35107385 on Build GRCh38
rs932599294 Chr.17: 35107325-35107325 on Build GRCh38
rs368838910 Chr.17: 35107397-35107397 on Build GRCh38
rs768717805 Chr.17: 35107433-35107433 on Build GRCh38
rs752679190 Chr.17: 35107479-35107479 on Build GRCh38
rs770147648 Chr.17: 35107418-35107418 on Build GRCh38
rs111671763 Chr.17: 35107390-35107390 on Build GRCh38
rs587780103 Chr.17: 35107382-35107382 on Build GRCh38
rs374986832 Chr.17: 35107488-35107488 on Build GRCh38
rs759935690 Chr.17: 35107326-35107326 on Build GRCh38
rs759532599 Chr.17: 35107455-35107455 on Build GRCh38
rs991317350 Chr.17: 35107294-35107294 on Build GRCh38
rs786203407 Chr.17: 35107379-35107379 on Build GRCh38
rs730881943 Chr.17: 35107440-35107441 on Build GRCh38
rs786202434 Chr.17: 35107380-35107381 on Build GRCh38
rs556011877 Chr.17: 35107497-35107497 on Build GRCh38
rs587782848 Chr.17: 35107376-35107376 on Build GRCh38
rs1024623836 Chr.17: 35107295-35107295 on Build GRCh38
rs750221187 Chr.17: 35107343-35107343 on Build GRCh38
rs56103933 Chr.17: 35107462-35107462 on Build GRCh38
rs539265370 Chr.17: 35107487-35107487 on Build GRCh38
rs764243244 Chr.17: 35107342-35107342 on Build GRCh38
rs765019908 Chr.17: 35107464-35107464 on Build GRCh38
rs876659926 Chr.17: 35107395-35107395 on Build GRCh38
rs753856224 Chr.17: 35107377-35107377 on Build GRCh38
rs370837197 Chr.17: 35107321-35107321 on Build GRCh38
rs771372627 Chr.17: 35107399-35107399 on Build GRCh38
rs766228268 Chr.17: 35107444-35107444 on Build GRCh38
rs730881946 Chr.17: 35107419-35107419 on Build GRCh38
rs775388851 Chr.17: 35107328-35107328 on Build GRCh38
rs753862052 Chr.17: 35107439-35107440 on Build GRCh38
rs786202507 Chr.17: 35107372-35107372 on Build GRCh38
rs876659394 Chr.17: 35107364-35107364 on Build GRCh38
rs375494170 Chr.17: 35107334-35107334 on Build GRCh38
rs762951311 Chr.17: 35107425-35107425 on Build GRCh38
rs142722898 Chr.17: 35107333-35107333 on Build GRCh38
rs781378161 Chr.17: 35107404-35107404 on Build GRCh38
rs760529001 Chr.17: 35107349-35107349 on Build GRCh38
rs1050979254 Chr.17: 35107340-35107340 on Build GRCh38
rs547475238 Chr.17: 35107436-35107436 on Build GRCh38
rs369396909 Chr.17: 35107378-35107378 on Build GRCh38
rs766067541 Chr.17: 35107351-35107351 on Build GRCh38
rs371561526 Chr.17: 35107443-35107443 on Build GRCh38

View Full Product Details