Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 136338175-136338446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAG1
Entrez Gene ID
10274
Gene Name
stromal antigen 1
Gene Aliases
SA1, SCC3A
UniGene
Hs.412586
Gene Chromosome Location
Chr.3: 136336235-136752412 on Build GRCh38
Transcripts
NM_005862.2 XM_011512329.2 XM_011512331.2 XM_017005523.1 XM_017005524.1 XM_017005525.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6379959 c.3698G>T p.R1233I Chr.3: 136338425-136338425 on Build GRCh38
6401575 c.3691C>T p.R1231* Chr.3: 136338432-136338432 on Build GRCh38
4433220 c.3736A>G p.I1246V Chr.3: 136338387-136338387 on Build GRCh38
1419365 c.3701C>T p.A1234V Chr.3: 136338422-136338422 on Build GRCh38
6227885 c.3731C>T p.A1244V Chr.3: 136338392-136338392 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777079352 Chr.3: 136338298-136338298 on Build GRCh38
rs374066787 Chr.3: 136338230-136338230 on Build GRCh38
rs374505679 Chr.3: 136338326-136338326 on Build GRCh38
rs375785966 Chr.3: 136338360-136338360 on Build GRCh38
rs772241339 Chr.3: 136338235-136338235 on Build GRCh38
rs770160031 Chr.3: 136338308-136338308 on Build GRCh38
rs754834612 Chr.3: 136338304-136338307 on Build GRCh38
rs749528012 Chr.3: 136338347-136338349 on Build GRCh38
rs758401374 Chr.3: 136338393-136338393 on Build GRCh38
rs747100449 Chr.3: 136338409-136338409 on Build GRCh38
rs771143295 Chr.3: 136338354-136338355 on Build GRCh38
rs749911602 Chr.3: 136338286-136338288 on Build GRCh38
rs374645509 Chr.3: 136338305-136338307 on Build GRCh38
rs773795522 Chr.3: 136338311-136338311 on Build GRCh38
rs769130382 Chr.3: 136338296-136338296 on Build GRCh38
rs145145175 Chr.3: 136338316-136338316 on Build GRCh38
rs111861599 Chr.3: 136338301-136338303 on Build GRCh38
rs763737876 Chr.3: 136338335-136338335 on Build GRCh38
rs764677928 Chr.3: 136338221-136338222 on Build GRCh38
rs199901285 Chr.3: 136338221-136338221 on Build GRCh38
rs761467929 Chr.3: 136338220-136338221 on Build GRCh38
rs755875634 Chr.3: 136338217-136338217 on Build GRCh38
rs765062646 Chr.3: 136338373-136338373 on Build GRCh38
rs762243866 Chr.3: 136338299-136338299 on Build GRCh38
rs188039314 Chr.3: 136338243-136338243 on Build GRCh38
rs750414067 Chr.3: 136338381-136338381 on Build GRCh38
rs747300515 Chr.3: 136338262-136338262 on Build GRCh38
rs192497288 Chr.3: 136338397-136338397 on Build GRCh38
rs376501222 Chr.3: 136338362-136338362 on Build GRCh38
rs533688925 Chr.3: 136338327-136338327 on Build GRCh38
rs757819960 Chr.3: 136338298-136338299 on Build GRCh38
rs369504607 Chr.3: 136338223-136338223 on Build GRCh38
rs376156087 Chr.3: 136338232-136338232 on Build GRCh38
rs954761736 Chr.3: 136338222-136338222 on Build GRCh38
rs780899926 Chr.3: 136338318-136338319 on Build GRCh38
rs767112064 Chr.3: 136338207-136338207 on Build GRCh38
rs757098419 Chr.3: 136338372-136338372 on Build GRCh38
rs763644751 Chr.3: 136338424-136338424 on Build GRCh38
rs751485802 Chr.3: 136338304-136338304 on Build GRCh38
rs745886329 Chr.3: 136338234-136338234 on Build GRCh38
rs182497205 Chr.3: 136338212-136338212 on Build GRCh38
rs760261047 Chr.3: 136338331-136338331 on Build GRCh38

View Full Product Details