Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47130886-47131121 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAE1
Entrez Gene ID
10055
Gene Name
SUMO1 activating enzyme subunit 1
Gene Aliases
AOS1, HSPC140, SUA1, UBLE1A
UniGene
Hs.515500
Gene Chromosome Location
Chr.19: 47130782-47210636 on Build GRCh38
Transcripts
NM_001145713.1 NM_001145714.1 NM_005500.2 NR_027280.1 XM_006722963.2 XM_006722965.3 XM_017026135.1 XM_017026136.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3823487 c.37G>C p.E13Q Chr.19: 47130967-47130967 on Build GRCh38
998677 c.93G>T p.Q31H Chr.19: 47131023-47131023 on Build GRCh38
6422210 c.45G>T p.E15D Chr.19: 47130975-47130975 on Build GRCh38
7316293 c.90C>T p.A30A Chr.19: 47131020-47131020 on Build GRCh38
7316294 c.90C>T p.A30A Chr.19: 47131020-47131020 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs895407920 Chr.19: 47130990-47130990 on Build GRCh38
rs177918 Chr.19: 47131056-47131056 on Build GRCh38
rs78321286 Chr.19: 47131091-47131091 on Build GRCh38
rs537729861 Chr.19: 47131086-47131086 on Build GRCh38
rs368510819 Chr.19: 47131031-47131031 on Build GRCh38
rs770492448 Chr.19: 47131043-47131043 on Build GRCh38
rs748761418 Chr.19: 47131035-47131035 on Build GRCh38
rs1008782457 Chr.19: 47131095-47131095 on Build GRCh38
rs376086221 Chr.19: 47130902-47130902 on Build GRCh38
rs538997359 Chr.19: 47130928-47130928 on Build GRCh38
rs879743914 Chr.19: 47131055-47131055 on Build GRCh38
rs557648651 Chr.19: 47131093-47131093 on Build GRCh38
rs370745297 Chr.19: 47130954-47130954 on Build GRCh38
rs753290677 Chr.19: 47130909-47130910 on Build GRCh38
rs747294644 Chr.19: 47130947-47130947 on Build GRCh38
rs968560874 Chr.19: 47131061-47131061 on Build GRCh38
rs1036098689 Chr.19: 47130939-47130939 on Build GRCh38
rs773977877 Chr.19: 47131054-47131054 on Build GRCh38
rs995923406 Chr.19: 47131078-47131082 on Build GRCh38
rs1037336406 Chr.19: 47131044-47131044 on Build GRCh38
rs868063534 Chr.19: 47130903-47130903 on Build GRCh38
rs1021127017 Chr.19: 47131097-47131097 on Build GRCh38
rs371247946 Chr.19: 47130942-47130942 on Build GRCh38
rs770360566 Chr.19: 47130984-47130984 on Build GRCh38
rs906517490 Chr.19: 47131037-47131037 on Build GRCh38
rs11670947 Chr.19: 47131051-47131051 on Build GRCh38
rs778739201 Chr.19: 47131088-47131088 on Build GRCh38
rs911537251 Chr.19: 47130924-47130924 on Build GRCh38
rs867127447 Chr.19: 47130908-47130908 on Build GRCh38
rs535230794 Chr.19: 47131017-47131017 on Build GRCh38
rs754472831 Chr.19: 47130952-47130957 on Build GRCh38
rs374154722 Chr.19: 47130957-47130957 on Build GRCh38
rs748567448 Chr.19: 47130972-47130972 on Build GRCh38
rs1030107045 Chr.19: 47131085-47131085 on Build GRCh38

View Full Product Details