Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 43218068-43218341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCUBE1
Entrez Gene ID
80274
Gene Name
signal peptide, CUB domain and EGF like domain containing 1
Gene Aliases
-
UniGene
Hs.133995
Gene Chromosome Location
Chr.22: 43203223-43343388 on Build GRCh38
Transcripts
NM_173050.3 XM_005261752.1 XM_011530389.2 XM_011530390.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4002198 c.1836G>C p.A612A Chr.22: 43218310-43218310 on Build GRCh38
1224986 c.1811A>G p.E604G Chr.22: 43218335-43218335 on Build GRCh38
6022040 c.1835C>T p.A612V Chr.22: 43218311-43218311 on Build GRCh38
6773504 c.1809C>T p.Y603Y Chr.22: 43218337-43218337 on Build GRCh38
6773503 c.1853C>T p.A618V Chr.22: 43218293-43218293 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779751816 Chr.22: 43218231-43218231 on Build GRCh38
rs752785457 Chr.22: 43218273-43218273 on Build GRCh38
rs746555989 Chr.22: 43218240-43218240 on Build GRCh38
rs1017376675 Chr.22: 43218124-43218124 on Build GRCh38
rs759584135 Chr.22: 43218269-43218269 on Build GRCh38
rs756587875 Chr.22: 43218204-43218204 on Build GRCh38
rs997774952 Chr.22: 43218160-43218160 on Build GRCh38
rs372224607 Chr.22: 43218222-43218222 on Build GRCh38
rs9623782 Chr.22: 43218217-43218217 on Build GRCh38
rs549148827 Chr.22: 43218205-43218205 on Build GRCh38
rs754221928 Chr.22: 43218280-43218280 on Build GRCh38
rs1031547012 Chr.22: 43218175-43218175 on Build GRCh38
rs570310016 Chr.22: 43218256-43218256 on Build GRCh38
rs182704661 Chr.22: 43218092-43218092 on Build GRCh38
rs9620123 Chr.22: 43218310-43218310 on Build GRCh38
rs769673353 Chr.22: 43218246-43218246 on Build GRCh38
rs998921659 Chr.22: 43218229-43218229 on Build GRCh38
rs376330838 Chr.22: 43218264-43218264 on Build GRCh38
rs781145321 Chr.22: 43218242-43218242 on Build GRCh38
rs553781614 Chr.22: 43218116-43218118 on Build GRCh38
rs764626411 Chr.22: 43218279-43218279 on Build GRCh38
rs751908189 Chr.22: 43218156-43218156 on Build GRCh38
rs754582888 Chr.22: 43218309-43218309 on Build GRCh38
rs947292127 Chr.22: 43218211-43218211 on Build GRCh38
rs752238746 Chr.22: 43218132-43218132 on Build GRCh38
rs369304150 Chr.22: 43218232-43218232 on Build GRCh38
rs970119576 Chr.22: 43218214-43218214 on Build GRCh38
rs768128524 Chr.22: 43218241-43218241 on Build GRCh38
rs749848924 Chr.22: 43218218-43218219 on Build GRCh38
rs754172036 Chr.22: 43218208-43218208 on Build GRCh38
rs952993042 Chr.22: 43218304-43218304 on Build GRCh38
rs56210050 Chr.22: 43218197-43218197 on Build GRCh38
rs749095995 Chr.22: 43218317-43218317 on Build GRCh38
rs112239566 Chr.22: 43218286-43218286 on Build GRCh38
rs199887506 Chr.22: 43218248-43218248 on Build GRCh38
rs576659990 Chr.22: 43218176-43218176 on Build GRCh38
rs748735874 Chr.22: 43218139-43218140 on Build GRCh38
rs767676628 Chr.22: 43218270-43218270 on Build GRCh38
rs12053796 Chr.22: 43218093-43218093 on Build GRCh38
rs1007286937 Chr.22: 43218099-43218099 on Build GRCh38
rs940011524 Chr.22: 43218101-43218101 on Build GRCh38
rs372406116 Chr.22: 43218311-43218311 on Build GRCh38
rs528749778 Chr.22: 43218244-43218244 on Build GRCh38
rs1028460278 Chr.22: 43218302-43218302 on Build GRCh38
rs761276482 Chr.22: 43218274-43218274 on Build GRCh38
rs779618810 Chr.22: 43218285-43218285 on Build GRCh38
rs563438727 Chr.22: 43218103-43218103 on Build GRCh38
rs1055528690 Chr.22: 43218193-43218193 on Build GRCh38

View Full Product Details