Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 132225588-132225861 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STK32C
Entrez Gene ID
282974
Gene Name
serine/threonine kinase 32C
Gene Aliases
PKE, YANK3
UniGene
Hs.469002
Gene Chromosome Location
Chr.10: 132207482-132332559 on Build GRCh38
Transcripts
NM_001318878.1 NM_001318879.1 NM_173575.3 NR_134911.1 XM_011539688.1 XM_011539690.2 XM_011539693.2 XM_011539694.1 XM_011539695.1 XM_011539696.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7234282 c.682+9G>A p.? Chr.10: 132225738-132225738 on Build GRCh38
4889398 c.726T>C p.H242H Chr.10: 132225612-132225612 on Build GRCh38
7234283 c.721+9G>A p.? Chr.10: 132225738-132225738 on Build GRCh38
4889399 c.687T>C p.H229H Chr.10: 132225612-132225612 on Build GRCh38
5796027 c.715G>A p.E239K Chr.10: 132225753-132225753 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371408061 Chr.10: 132225633-132225633 on Build GRCh38
rs368143296 Chr.10: 132225667-132225667 on Build GRCh38
rs766518082 Chr.10: 132225726-132225726 on Build GRCh38
rs745602506 Chr.10: 132225792-132225792 on Build GRCh38
rs759846989 Chr.10: 132225818-132225818 on Build GRCh38
rs754037302 Chr.10: 132225737-132225737 on Build GRCh38
rs374665854 Chr.10: 132225666-132225666 on Build GRCh38
rs759334059 Chr.10: 132225620-132225620 on Build GRCh38
rs79967518 Chr.10: 132225807-132225807 on Build GRCh38
rs758767447 Chr.10: 132225828-132225828 on Build GRCh38
rs769299549 Chr.10: 132225794-132225794 on Build GRCh38
rs918840872 Chr.10: 132225681-132225681 on Build GRCh38
rs942731294 Chr.10: 132225719-132225719 on Build GRCh38
rs371783935 Chr.10: 132225699-132225699 on Build GRCh38
rs370952269 Chr.10: 132225690-132225690 on Build GRCh38
rs555980878 Chr.10: 132225802-132225802 on Build GRCh38
rs2130893 Chr.10: 132225628-132225628 on Build GRCh38
rs902424973 Chr.10: 132225816-132225816 on Build GRCh38
rs576266657 Chr.10: 132225755-132225755 on Build GRCh38
rs867019286 Chr.10: 132225805-132225805 on Build GRCh38
rs764710256 Chr.10: 132225835-132225835 on Build GRCh38
rs573919284 Chr.10: 132225660-132225660 on Build GRCh38
rs753142771 Chr.10: 132225825-132225825 on Build GRCh38
rs201250732 Chr.10: 132225626-132225626 on Build GRCh38
rs150199741 Chr.10: 132225743-132225743 on Build GRCh38
rs540990409 Chr.10: 132225809-132225809 on Build GRCh38
rs767752738 Chr.10: 132225827-132225828 on Build GRCh38
rs780276302 Chr.10: 132225631-132225631 on Build GRCh38
rs778382575 Chr.10: 132225757-132225757 on Build GRCh38
rs372041069 Chr.10: 132225665-132225665 on Build GRCh38
rs766542843 Chr.10: 132225815-132225815 on Build GRCh38
rs765326155 Chr.10: 132225819-132225819 on Build GRCh38
rs187244260 Chr.10: 132225670-132225670 on Build GRCh38
rs769689233 Chr.10: 132225664-132225664 on Build GRCh38
rs774210431 Chr.10: 132225808-132225808 on Build GRCh38
rs760708692 Chr.10: 132225701-132225701 on Build GRCh38
rs746255593 Chr.10: 132225629-132225629 on Build GRCh38
rs780344299 Chr.10: 132225722-132225722 on Build GRCh38
rs746467023 Chr.10: 132225650-132225650 on Build GRCh38
rs753087591 Chr.10: 132225751-132225751 on Build GRCh38
rs200776513 Chr.10: 132225657-132225657 on Build GRCh38
rs776538654 Chr.10: 132225817-132225817 on Build GRCh38
rs145857222 Chr.10: 132225766-132225766 on Build GRCh38
rs549305020 Chr.10: 132225647-132225647 on Build GRCh38
rs748752599 Chr.10: 132225644-132225644 on Build GRCh38
rs369505380 Chr.10: 132225671-132225671 on Build GRCh38
rs772671489 Chr.10: 132225645-132225645 on Build GRCh38
rs529188501 Chr.10: 132225638-132225638 on Build GRCh38
rs370902327 Chr.10: 132225669-132225669 on Build GRCh38
rs7916962 Chr.10: 132225659-132225659 on Build GRCh38
rs939839027 Chr.10: 132225698-132225698 on Build GRCh38
rs775406700 Chr.10: 132225798-132225798 on Build GRCh38
rs2130894 Chr.10: 132225738-132225738 on Build GRCh38
rs1009650369 Chr.10: 132225636-132225636 on Build GRCh38
rs909746687 Chr.10: 132225694-132225694 on Build GRCh38
rs900296721 Chr.10: 132225773-132225773 on Build GRCh38
rs1048369388 Chr.10: 132225693-132225693 on Build GRCh38
rs137968970 Chr.10: 132225818-132225819 on Build GRCh38
rs780553581 Chr.10: 132225788-132225788 on Build GRCh38
rs756769561 Chr.10: 132225780-132225780 on Build GRCh38

View Full Product Details