Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8253857-8254098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PFAS
Entrez Gene ID
5198
Gene Name
phosphoribosylformylglycinamidine synthase
Gene Aliases
FGAMS, FGAR-AT, FGARAT, PURL
UniGene
Hs.573976
Gene Chromosome Location
Chr.17: 8249255-8270491 on Build GRCh38
Transcripts
NM_012393.2 XM_006721546.2 XM_017024760.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1387635 c.19T>G p.F7V Chr.17: 8253956-8253956 on Build GRCh38
4504971 c.67C>T p.R23W Chr.17: 8254004-8254004 on Build GRCh38
7173958 c.115G>A p.E39K Chr.17: 8254052-8254052 on Build GRCh38
6250846 c.142+6G>T p.? Chr.17: 8254085-8254085 on Build GRCh38
2882216 c.39C>T p.G13G Chr.17: 8253976-8253976 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754133256 Chr.17: 8253963-8253963 on Build GRCh38
rs758832526 Chr.17: 8253981-8253981 on Build GRCh38
rs747899881 Chr.17: 8253994-8253994 on Build GRCh38
rs776708331 Chr.17: 8254023-8254023 on Build GRCh38
rs967909286 Chr.17: 8253892-8253892 on Build GRCh38
rs755291903 Chr.17: 8254061-8254061 on Build GRCh38
rs748839729 Chr.17: 8254004-8254004 on Build GRCh38
rs1048606410 Chr.17: 8253998-8253998 on Build GRCh38
rs374748339 Chr.17: 8253910-8253910 on Build GRCh38
rs188398517 Chr.17: 8253925-8253925 on Build GRCh38
rs28523368 Chr.17: 8253890-8253890 on Build GRCh38
rs369136545 Chr.17: 8254078-8254078 on Build GRCh38
rs573066925 Chr.17: 8254005-8254005 on Build GRCh38
rs974202184 Chr.17: 8254017-8254017 on Build GRCh38
rs368050168 Chr.17: 8253982-8253982 on Build GRCh38
rs762041987 Chr.17: 8254025-8254025 on Build GRCh38
rs749327793 Chr.17: 8253890-8253891 on Build GRCh38
rs763042045 Chr.17: 8253932-8253932 on Build GRCh38
rs371924315 Chr.17: 8254066-8254066 on Build GRCh38
rs150944821 Chr.17: 8253970-8253970 on Build GRCh38
rs192637030 Chr.17: 8253950-8253950 on Build GRCh38
rs750458468 Chr.17: 8254042-8254042 on Build GRCh38
rs773017916 Chr.17: 8253929-8253929 on Build GRCh38
rs765377606 Chr.17: 8254038-8254038 on Build GRCh38
rs752099825 Chr.17: 8254054-8254054 on Build GRCh38
rs765420234 Chr.17: 8253966-8253966 on Build GRCh38
rs528012391 Chr.17: 8253889-8253889 on Build GRCh38
rs9891699 Chr.17: 8253992-8253992 on Build GRCh38
rs766846644 Chr.17: 8254046-8254046 on Build GRCh38
rs776366752 Chr.17: 8253922-8253922 on Build GRCh38
rs748475399 Chr.17: 8254067-8254067 on Build GRCh38
rs759376718 Chr.17: 8254009-8254009 on Build GRCh38
rs769172804 Chr.17: 8253996-8253996 on Build GRCh38
rs893279600 Chr.17: 8253956-8253956 on Build GRCh38
rs772781776 Chr.17: 8254001-8254001 on Build GRCh38
rs368056582 Chr.17: 8253894-8253894 on Build GRCh38
rs368608162 Chr.17: 8253927-8253927 on Build GRCh38
rs930236952 Chr.17: 8254064-8254064 on Build GRCh38
rs183954034 Chr.17: 8253965-8253965 on Build GRCh38
rs200607158 Chr.17: 8253960-8253960 on Build GRCh38
rs920195590 Chr.17: 8254036-8254036 on Build GRCh38
rs759427892 Chr.17: 8253962-8253962 on Build GRCh38
rs755417531 Chr.17: 8253991-8253991 on Build GRCh38
rs767415963 Chr.17: 8254014-8254014 on Build GRCh38
rs774589479 Chr.17: 8254008-8254019 on Build GRCh38
rs761042116 Chr.17: 8254041-8254041 on Build GRCh38
rs771070491 Chr.17: 8253891-8253910 on Build GRCh38

View Full Product Details