Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33917079-33917340 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBE2R2
Entrez Gene ID
54926
Gene Name
ubiquitin conjugating enzyme E2 R2
Gene Aliases
CDC34B, E2-CDC34B, UBC3B
UniGene
Hs.643648
Gene Chromosome Location
Chr.9: 33817164-33920403 on Build GRCh38
Transcripts
NM_017811.3 XM_005251496.2 XM_011517949.1 XM_017014860.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5628354 c.642_643insA p.I215fs*2 Chr.9: 33917162-33917163 on Build GRCh38
5926705 c.661G>A p.E221K Chr.9: 33917181-33917181 on Build GRCh38
1319606 c.658_660delGAG p.E223delE Chr.9: 33917178-33917180 on Build GRCh38
6815162 c.713C>T p.S238L Chr.9: 33917233-33917233 on Build GRCh38
2772898 c.564G>A p.A188A Chr.9: 33917084-33917084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143620598 Chr.9: 33917160-33917160 on Build GRCh38
rs370067900 Chr.9: 33917234-33917234 on Build GRCh38
rs750567309 Chr.9: 33917141-33917141 on Build GRCh38
rs199597685 Chr.9: 33917180-33917180 on Build GRCh38
rs762050649 Chr.9: 33917251-33917251 on Build GRCh38
rs778731566 Chr.9: 33917177-33917177 on Build GRCh38
rs763320246 Chr.9: 33917116-33917116 on Build GRCh38
rs970490787 Chr.9: 33917244-33917253 on Build GRCh38
rs775980700 Chr.9: 33917238-33917238 on Build GRCh38
rs1026908813 Chr.9: 33917194-33917194 on Build GRCh38
rs187812808 Chr.9: 33917242-33917242 on Build GRCh38
rs367990862 Chr.9: 33917204-33917204 on Build GRCh38
rs960341662 Chr.9: 33917233-33917233 on Build GRCh38
rs767843659 Chr.9: 33917261-33917261 on Build GRCh38
rs764452731 Chr.9: 33917117-33917117 on Build GRCh38
rs760246701 Chr.9: 33917210-33917210 on Build GRCh38
rs148897042 Chr.9: 33917159-33917159 on Build GRCh38
rs1044452 Chr.9: 33917306-33917306 on Build GRCh38
rs753745524 Chr.9: 33917172-33917174 on Build GRCh38
rs752002054 Chr.9: 33917245-33917245 on Build GRCh38
rs1040266366 Chr.9: 33917304-33917304 on Build GRCh38
rs867080039 Chr.9: 33917221-33917221 on Build GRCh38
rs371982376 Chr.9: 33917271-33917271 on Build GRCh38
rs772466935 Chr.9: 33917178-33917180 on Build GRCh38
rs771807900 Chr.9: 33917196-33917196 on Build GRCh38
rs372565481 Chr.9: 33917123-33917123 on Build GRCh38
rs943206337 Chr.9: 33917298-33917298 on Build GRCh38
rs184815327 Chr.9: 33917156-33917156 on Build GRCh38
rs145589627 Chr.9: 33917130-33917130 on Build GRCh38
rs756272723 Chr.9: 33917281-33917281 on Build GRCh38
rs756254530 Chr.9: 33917144-33917144 on Build GRCh38
rs376151379 Chr.9: 33917133-33917133 on Build GRCh38
rs959035413 Chr.9: 33917315-33917315 on Build GRCh38
rs764184049 Chr.9: 33917203-33917205 on Build GRCh38
rs763409507 Chr.9: 33917239-33917239 on Build GRCh38

View Full Product Details