Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 23047595-23047868 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THBD
Entrez Gene ID
7056
Gene Name
thrombomodulin
Gene Aliases
AHUS6, BDCA3, CD141, THPH12, THRM, TM
UniGene
Hs.2030
Gene Chromosome Location
Chr.20: 23045633-23049664 on Build GRCh38
Transcripts
NM_000361.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6801075 c.1663G>A p.A555T Chr.20: 23047842-23047842 on Build GRCh38
6801084 c.1700G>A p.R567Q Chr.20: 23047805-23047805 on Build GRCh38
183315 c.1664C>T p.A555V Chr.20: 23047841-23047841 on Build GRCh38
4837622 c.1670C>A p.P557H Chr.20: 23047835-23047835 on Build GRCh38
1307209 c.1721G>A p.R574K Chr.20: 23047784-23047784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs974282704 Chr.20: 23047791-23047791 on Build GRCh38
rs542603082 Chr.20: 23047664-23047664 on Build GRCh38
rs368774068 Chr.20: 23047805-23047805 on Build GRCh38
rs539089204 Chr.20: 23047782-23047782 on Build GRCh38
rs559059986 Chr.20: 23047667-23047667 on Build GRCh38
rs778136712 Chr.20: 23047833-23047833 on Build GRCh38
rs777258948 Chr.20: 23047846-23047846 on Build GRCh38
rs763321484 Chr.20: 23047781-23047781 on Build GRCh38
rs868105152 Chr.20: 23047784-23047784 on Build GRCh38
rs3176121 Chr.20: 23047619-23047619 on Build GRCh38
rs202143136 Chr.20: 23047792-23047792 on Build GRCh38
rs775103380 Chr.20: 23047734-23047734 on Build GRCh38
rs201487514 Chr.20: 23047827-23047827 on Build GRCh38
rs763887670 Chr.20: 23047798-23047798 on Build GRCh38
rs751317027 Chr.20: 23047801-23047801 on Build GRCh38
rs747367882 Chr.20: 23047834-23047834 on Build GRCh38
rs903959463 Chr.20: 23047843-23047843 on Build GRCh38
rs779607352 Chr.20: 23047812-23047812 on Build GRCh38
rs572561997 Chr.20: 23047668-23047668 on Build GRCh38
rs553113676 Chr.20: 23047626-23047626 on Build GRCh38
rs750407288 Chr.20: 23047806-23047806 on Build GRCh38
rs950398067 Chr.20: 23047761-23047761 on Build GRCh38
rs765445275 Chr.20: 23047768-23047768 on Build GRCh38
rs377311614 Chr.20: 23047736-23047736 on Build GRCh38
rs867709352 Chr.20: 23047842-23047842 on Build GRCh38
rs143625916 Chr.20: 23047836-23047836 on Build GRCh38
rs376256445 Chr.20: 23047661-23047661 on Build GRCh38
rs13306851 Chr.20: 23047767-23047767 on Build GRCh38
rs773693706 Chr.20: 23047637-23047637 on Build GRCh38
rs908965996 Chr.20: 23047773-23047773 on Build GRCh38
rs141651047 Chr.20: 23047702-23047702 on Build GRCh38
rs752959310 Chr.20: 23047774-23047774 on Build GRCh38
rs983113093 Chr.20: 23047769-23047769 on Build GRCh38
rs760823569 Chr.20: 23047755-23047772 on Build GRCh38
rs41328344 Chr.20: 23047741-23047741 on Build GRCh38
rs912809279 Chr.20: 23047809-23047809 on Build GRCh38
rs891152417 Chr.20: 23047677-23047677 on Build GRCh38
rs936532361 Chr.20: 23047659-23047659 on Build GRCh38
rs868614791 Chr.20: 23047790-23047790 on Build GRCh38
rs761189965 Chr.20: 23047746-23047746 on Build GRCh38
rs774607361 Chr.20: 23047779-23047779 on Build GRCh38
rs566980627 Chr.20: 23047757-23047757 on Build GRCh38
rs565111682 Chr.20: 23047715-23047715 on Build GRCh38
rs865948837 Chr.20: 23047800-23047800 on Build GRCh38
rs13306852 Chr.20: 23047751-23047751 on Build GRCh38
rs1051122432 Chr.20: 23047671-23047671 on Build GRCh38
rs573584085 Chr.20: 23047630-23047630 on Build GRCh38
rs576345574 Chr.20: 23047737-23047754 on Build GRCh38
rs932869999 Chr.20: 23047660-23047660 on Build GRCh38
rs3176133 Chr.20: 23047690-23047690 on Build GRCh38
rs530087674 Chr.20: 23047750-23047750 on Build GRCh38
rs77670413 Chr.20: 23047726-23047726 on Build GRCh38
rs551028498 Chr.20: 23047730-23047730 on Build GRCh38
rs1000711528 Chr.20: 23047733-23047733 on Build GRCh38
rs755593080 Chr.20: 23047808-23047808 on Build GRCh38

View Full Product Details