Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55376805-55377078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM190
Entrez Gene ID
147744
Gene Name
transmembrane protein 190
Gene Aliases
MDAC1
UniGene
Hs.590943
Gene Chromosome Location
Chr.19: 55376126-55378246 on Build GRCh38
Transcripts
NM_139172.1 XM_017026331.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3960537 c.81C>T p.F27F Chr.19: 55377013-55377013 on Build GRCh38
128662 c.54_55delGG p.A19fs*15 Chr.19: 55376907-55376908 on Build GRCh38
5761182 c.60C>T p.D20D Chr.19: 55376992-55376992 on Build GRCh38
5390810 c.77G>A p.G26E Chr.19: 55377009-55377009 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs932851543 Chr.19: 55377032-55377032 on Build GRCh38
rs1016058051 Chr.19: 55376931-55376931 on Build GRCh38
rs548963377 Chr.19: 55376930-55376930 on Build GRCh38
rs33999165 Chr.19: 55377025-55377026 on Build GRCh38
rs762707103 Chr.19: 55377022-55377022 on Build GRCh38
rs757148152 Chr.19: 55376935-55376935 on Build GRCh38
rs762690432 Chr.19: 55376928-55376928 on Build GRCh38
rs777272573 Chr.19: 55376939-55376941 on Build GRCh38
rs763752952 Chr.19: 55376929-55376929 on Build GRCh38
rs1014645659 Chr.19: 55376939-55376939 on Build GRCh38
rs1003171235 Chr.19: 55376851-55376851 on Build GRCh38
rs976941680 Chr.19: 55377014-55377014 on Build GRCh38
rs751154914 Chr.19: 55376940-55376941 on Build GRCh38
rs767450734 Chr.19: 55376937-55376937 on Build GRCh38
rs757868196 Chr.19: 55376907-55376917 on Build GRCh38
rs752437621 Chr.19: 55376913-55376913 on Build GRCh38
rs746213919 Chr.19: 55376987-55376988 on Build GRCh38
rs779797713 Chr.19: 55376950-55376950 on Build GRCh38
rs749493487 Chr.19: 55376842-55376842 on Build GRCh38
rs773478209 Chr.19: 55376877-55376877 on Build GRCh38
rs768768677 Chr.19: 55376867-55376867 on Build GRCh38
rs199617765 Chr.19: 55376952-55376952 on Build GRCh38
rs763773512 Chr.19: 55377031-55377031 on Build GRCh38
rs771489264 Chr.19: 55376992-55376992 on Build GRCh38
rs766915709 Chr.19: 55377044-55377044 on Build GRCh38
rs201520200 Chr.19: 55376993-55376993 on Build GRCh38
rs903104944 Chr.19: 55376835-55376835 on Build GRCh38
rs780776383 Chr.19: 55376948-55376953 on Build GRCh38
rs558068266 Chr.19: 55376963-55376963 on Build GRCh38
rs956364162 Chr.19: 55376986-55376986 on Build GRCh38
rs750208291 Chr.19: 55376943-55376943 on Build GRCh38
rs1038984233 Chr.19: 55376833-55376833 on Build GRCh38
rs756889353 Chr.19: 55376944-55376945 on Build GRCh38
rs769940682 Chr.19: 55376838-55376838 on Build GRCh38
rs535160596 Chr.19: 55376876-55376876 on Build GRCh38
rs772476710 Chr.19: 55376965-55376965 on Build GRCh38
rs747439304 Chr.19: 55376987-55376987 on Build GRCh38
rs944407916 Chr.19: 55377019-55377019 on Build GRCh38
rs537247338 Chr.19: 55377037-55377037 on Build GRCh38
rs369802467 Chr.19: 55377036-55377036 on Build GRCh38
rs774412761 Chr.19: 55376874-55376874 on Build GRCh38
rs982508405 Chr.19: 55376981-55376981 on Build GRCh38
rs755182862 Chr.19: 55376953-55376953 on Build GRCh38
rs565648905 Chr.19: 55376948-55376948 on Build GRCh38
rs766824649 Chr.19: 55376886-55376886 on Build GRCh38
rs575331308 Chr.19: 55377051-55377051 on Build GRCh38
rs778875921 Chr.19: 55376961-55376961 on Build GRCh38

View Full Product Details