Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90959833-90960066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RCCD1
Entrez Gene ID
91433
Gene Name
RCC1 domain containing 1
Gene Aliases
-
UniGene
Hs.655895
Gene Chromosome Location
Chr.15: 90954864-90964858 on Build GRCh38
Transcripts
NM_001017919.1 NM_033544.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6425178 c.710A>C p.N237T Chr.15: 90959930-90959930 on Build GRCh38
6001601 c.723G>A p.Q241Q Chr.15: 90959943-90959943 on Build GRCh38
6393093 c.719G>T p.G240V Chr.15: 90959939-90959939 on Build GRCh38
6410040 c.741G>T p.R247S Chr.15: 90959961-90959961 on Build GRCh38
6422576 c.772A>T p.R258W Chr.15: 90959992-90959992 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770136569 Chr.15: 90959900-90959900 on Build GRCh38
rs201626365 Chr.15: 90959951-90959951 on Build GRCh38
rs770010766 Chr.15: 90960013-90960013 on Build GRCh38
rs749077411 Chr.15: 90959854-90959854 on Build GRCh38
rs747944046 Chr.15: 90959989-90959989 on Build GRCh38
rs745985461 Chr.15: 90959889-90959889 on Build GRCh38
rs1040703588 Chr.15: 90959901-90959901 on Build GRCh38
rs538994201 Chr.15: 90960008-90960008 on Build GRCh38
rs148056787 Chr.15: 90959988-90959988 on Build GRCh38
rs1007498037 Chr.15: 90959998-90959999 on Build GRCh38
rs527313952 Chr.15: 90959963-90959963 on Build GRCh38
rs777925352 Chr.15: 90959993-90959993 on Build GRCh38
rs767674717 Chr.15: 90959927-90959927 on Build GRCh38
rs111371919 Chr.15: 90959885-90959885 on Build GRCh38
rs760120190 Chr.15: 90960036-90960036 on Build GRCh38
rs373420827 Chr.15: 90960030-90960030 on Build GRCh38
rs369168110 Chr.15: 90960026-90960026 on Build GRCh38
rs758847257 Chr.15: 90959958-90959958 on Build GRCh38
rs758327549 Chr.15: 90959990-90959990 on Build GRCh38
rs111477163 Chr.15: 90959883-90959883 on Build GRCh38
rs753447545 Chr.15: 90960037-90960037 on Build GRCh38
rs756880663 Chr.15: 90959971-90959973 on Build GRCh38
rs1037812535 Chr.15: 90959944-90959944 on Build GRCh38
rs569388463 Chr.15: 90960039-90960039 on Build GRCh38
rs773765473 Chr.15: 90960031-90960031 on Build GRCh38
rs780499670 Chr.15: 90959983-90959983 on Build GRCh38
rs139470494 Chr.15: 90960012-90960012 on Build GRCh38
rs75390535 Chr.15: 90959965-90959965 on Build GRCh38
rs775792152 Chr.15: 90960015-90960015 on Build GRCh38
rs779502043 Chr.15: 90959879-90959879 on Build GRCh38
rs757216509 Chr.15: 90960003-90960003 on Build GRCh38
rs978607942 Chr.15: 90960044-90960044 on Build GRCh38
rs767956222 Chr.15: 90959964-90959964 on Build GRCh38
rs369101925 Chr.15: 90959858-90959858 on Build GRCh38
rs775916698 Chr.15: 90959946-90959946 on Build GRCh38
rs763356428 Chr.15: 90959949-90959949 on Build GRCh38
rs373719530 Chr.15: 90960044-90960045 on Build GRCh38
rs745653171 Chr.15: 90960007-90960008 on Build GRCh38
rs753344520 Chr.15: 90959863-90959864 on Build GRCh38
rs1006815086 Chr.15: 90959919-90959919 on Build GRCh38
rs774683296 Chr.15: 90959959-90959959 on Build GRCh38
rs749559134 Chr.15: 90960025-90960025 on Build GRCh38
rs766001525 Chr.15: 90959975-90959975 on Build GRCh38
rs202026116 Chr.15: 90959991-90959991 on Build GRCh38

View Full Product Details