Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43131957-43132228 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTK7
Entrez Gene ID
5754
Gene Name
protein tyrosine kinase 7 (inactive)
Gene Aliases
CCK-4, CCK4
UniGene
Hs.90572
Gene Chromosome Location
Chr.6: 43076268-43161720 on Build GRCh38
Transcripts
NM_001270398.1 NM_002821.4 NM_152880.3 NM_152881.3 NM_152882.3 NR_072997.1 NR_072998.1 XM_011514765.2 XM_011514766.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3079823 c.826C>T p.R276C Chr.6: 43132029-43132029 on Build GRCh38
6504441 c.133C>T p.R45W Chr.6: 43132077-43132077 on Build GRCh38
1444645 c.837A>G p.T279T Chr.6: 43132040-43132040 on Build GRCh38
5430627 c.920G>A p.R307K Chr.6: 43132123-43132123 on Build GRCh38
6897852 c.839T>A p.V280E Chr.6: 43132042-43132042 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762869298 Chr.6: 43131990-43131990 on Build GRCh38
rs151126990 Chr.6: 43132021-43132021 on Build GRCh38
rs765804266 Chr.6: 43132022-43132022 on Build GRCh38
rs201871298 Chr.6: 43132184-43132184 on Build GRCh38
rs56188167 Chr.6: 43132030-43132030 on Build GRCh38
rs1043875969 Chr.6: 43132138-43132138 on Build GRCh38
rs752143475 Chr.6: 43132026-43132026 on Build GRCh38
rs946967711 Chr.6: 43132111-43132111 on Build GRCh38
rs750854100 Chr.6: 43132177-43132177 on Build GRCh38
rs564569746 Chr.6: 43132019-43132019 on Build GRCh38
rs552297655 Chr.6: 43132083-43132083 on Build GRCh38
rs147935332 Chr.6: 43132118-43132118 on Build GRCh38
rs767765958 Chr.6: 43132164-43132164 on Build GRCh38
rs372757537 Chr.6: 43131995-43131995 on Build GRCh38
rs749415736 Chr.6: 43132070-43132070 on Build GRCh38
rs774104483 Chr.6: 43131997-43131997 on Build GRCh38
rs374009568 Chr.6: 43132029-43132029 on Build GRCh38
rs752053533 Chr.6: 43132157-43132157 on Build GRCh38
rs773128495 Chr.6: 43132102-43132102 on Build GRCh38
rs768758723 Chr.6: 43132074-43132074 on Build GRCh38
rs759077094 Chr.6: 43132122-43132122 on Build GRCh38
rs767370598 Chr.6: 43132017-43132017 on Build GRCh38
rs775641713 Chr.6: 43131985-43131985 on Build GRCh38
rs376406517 Chr.6: 43132192-43132192 on Build GRCh38
rs774225529 Chr.6: 43132077-43132077 on Build GRCh38
rs540612548 Chr.6: 43132065-43132065 on Build GRCh38
rs141712145 Chr.6: 43132161-43132161 on Build GRCh38
rs779981129 Chr.6: 43132069-43132069 on Build GRCh38
rs55849103 Chr.6: 43132106-43132106 on Build GRCh38
rs902689635 Chr.6: 43132194-43132194 on Build GRCh38
rs753457277 Chr.6: 43132023-43132023 on Build GRCh38
rs764062621 Chr.6: 43132005-43132005 on Build GRCh38
rs761561036 Chr.6: 43132001-43132001 on Build GRCh38
rs375898751 Chr.6: 43132018-43132018 on Build GRCh38
rs748100799 Chr.6: 43132084-43132084 on Build GRCh38
rs764859448 Chr.6: 43132127-43132127 on Build GRCh38
rs200192410 Chr.6: 43132085-43132085 on Build GRCh38
rs144850065 Chr.6: 43132038-43132038 on Build GRCh38
rs141004962 Chr.6: 43132024-43132024 on Build GRCh38
rs934010105 Chr.6: 43132054-43132054 on Build GRCh38
rs892398402 Chr.6: 43132089-43132089 on Build GRCh38
rs766105075 Chr.6: 43132115-43132115 on Build GRCh38
rs778463968 Chr.6: 43132025-43132025 on Build GRCh38
rs199784710 Chr.6: 43132033-43132033 on Build GRCh38

View Full Product Details