Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 125076628-125076897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC37A2
Entrez Gene ID
219855
Gene Name
solute carrier family 37 member 2
Gene Aliases
pp11662
UniGene
Hs.352661
Gene Chromosome Location
Chr.11: 125063067-125090516 on Build GRCh38
Transcripts
NM_001145290.1 NM_198277.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5640613 c.65G>A p.R22Q Chr.11: 125076762-125076762 on Build GRCh38
1226325 c.64C>T p.R22* Chr.11: 125076761-125076761 on Build GRCh38
6068047 c.77T>A p.L26Q Chr.11: 125076774-125076774 on Build GRCh38
6068046 c.77T>A p.L26Q Chr.11: 125076774-125076774 on Build GRCh38
6131813 c.135C>A p.I45I Chr.11: 125076832-125076832 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570788891 Chr.11: 125076793-125076793 on Build GRCh38
rs188435391 Chr.11: 125076827-125076827 on Build GRCh38
rs145893905 Chr.11: 125076798-125076798 on Build GRCh38
rs148933416 Chr.11: 125076833-125076833 on Build GRCh38
rs760123981 Chr.11: 125076783-125076783 on Build GRCh38
rs750250353 Chr.11: 125076804-125076804 on Build GRCh38
rs758355318 Chr.11: 125076809-125076809 on Build GRCh38
rs61912691 Chr.11: 125076862-125076862 on Build GRCh38
rs78922475 Chr.11: 125076796-125076796 on Build GRCh38
rs373111916 Chr.11: 125076664-125076664 on Build GRCh38
rs776330729 Chr.11: 125076854-125076854 on Build GRCh38
rs568518370 Chr.11: 125076662-125076662 on Build GRCh38
rs772045331 Chr.11: 125076684-125076684 on Build GRCh38
rs760039229 Chr.11: 125076691-125076691 on Build GRCh38
rs748253509 Chr.11: 125076753-125076753 on Build GRCh38
rs761308782 Chr.11: 125076855-125076855 on Build GRCh38
rs966437031 Chr.11: 125076765-125076765 on Build GRCh38
rs571262164 Chr.11: 125076832-125076832 on Build GRCh38
rs73625344 Chr.11: 125076733-125076733 on Build GRCh38
rs374174542 Chr.11: 125076762-125076762 on Build GRCh38
rs138498389 Chr.11: 125076830-125076830 on Build GRCh38
rs763665609 Chr.11: 125076725-125076725 on Build GRCh38
rs772570321 Chr.11: 125076775-125076775 on Build GRCh38
rs779180483 Chr.11: 125076828-125076828 on Build GRCh38
rs746439252 Chr.11: 125076769-125076769 on Build GRCh38
rs781358402 Chr.11: 125076752-125076752 on Build GRCh38
rs752786146 Chr.11: 125076821-125076821 on Build GRCh38
rs771251345 Chr.11: 125076761-125076761 on Build GRCh38
rs112732048 Chr.11: 125076826-125076826 on Build GRCh38
rs775079404 Chr.11: 125076843-125076843 on Build GRCh38
rs768097551 Chr.11: 125076840-125076840 on Build GRCh38
rs753045059 Chr.11: 125076797-125076797 on Build GRCh38
rs866259250 Chr.11: 125076660-125076660 on Build GRCh38
rs753617224 Chr.11: 125076732-125076732 on Build GRCh38
rs548665564 Chr.11: 125076657-125076657 on Build GRCh38
rs766446225 Chr.11: 125076817-125076817 on Build GRCh38
rs749633539 Chr.11: 125076760-125076760 on Build GRCh38
rs750431954 Chr.11: 125076744-125076744 on Build GRCh38
rs1028499028 Chr.11: 125076704-125076704 on Build GRCh38
rs778034288 Chr.11: 125076824-125076824 on Build GRCh38
rs765231205 Chr.11: 125076734-125076734 on Build GRCh38
rs751829429 Chr.11: 125076682-125076682 on Build GRCh38
rs926206216 Chr.11: 125076724-125076724 on Build GRCh38
rs527877172 Chr.11: 125076681-125076681 on Build GRCh38
rs777890036 Chr.11: 125076755-125076755 on Build GRCh38
rs755108229 Chr.11: 125076746-125076746 on Build GRCh38
rs1008394178 Chr.11: 125076740-125076740 on Build GRCh38

View Full Product Details