Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 39837883-39838138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDS5A
Entrez Gene ID
23244
Gene Name
PDS5 cohesin associated factor A
Gene Aliases
PIG54, SCC-112, SCC112
UniGene
Hs.331431
Gene Chromosome Location
Chr.4: 39822863-39977956 on Build GRCh38
Transcripts
NM_001100399.1 XM_011513672.2 XM_017007928.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6167100 c.3782G>T p.G1261V Chr.4: 39838084-39838084 on Build GRCh38
3603596 c.3871C>T p.P1291S Chr.4: 39837995-39837995 on Build GRCh38
351020 c.3891G>C p.K1297N Chr.4: 39837975-39837975 on Build GRCh38
5908423 c.3793C>T p.P1265S Chr.4: 39838073-39838073 on Build GRCh38
2846781 c.3757_3758insA p.T1253fs*3 Chr.4: 39838108-39838109 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755186328 Chr.4: 39837944-39837944 on Build GRCh38
rs780111750 Chr.4: 39838108-39838108 on Build GRCh38
rs762301124 Chr.4: 39838062-39838062 on Build GRCh38
rs781408731 Chr.4: 39838059-39838059 on Build GRCh38
rs745736268 Chr.4: 39837976-39837976 on Build GRCh38
rs781346079 Chr.4: 39837971-39837971 on Build GRCh38
rs752859586 Chr.4: 39838078-39838078 on Build GRCh38
rs761119811 Chr.4: 39838070-39838070 on Build GRCh38
rs759590781 Chr.4: 39837938-39837938 on Build GRCh38
rs748927296 Chr.4: 39838049-39838049 on Build GRCh38
rs574475863 Chr.4: 39838116-39838116 on Build GRCh38
rs539362968 Chr.4: 39838053-39838053 on Build GRCh38
rs554644538 Chr.4: 39838104-39838104 on Build GRCh38
rs772352579 Chr.4: 39838063-39838063 on Build GRCh38
rs781080179 Chr.4: 39838086-39838086 on Build GRCh38
rs200621682 Chr.4: 39838087-39838087 on Build GRCh38
rs756186770 Chr.4: 39837935-39837935 on Build GRCh38
rs765553747 Chr.4: 39838077-39838077 on Build GRCh38
rs764522553 Chr.4: 39838075-39838075 on Build GRCh38
rs959314223 Chr.4: 39837987-39837987 on Build GRCh38
rs372585732 Chr.4: 39838072-39838072 on Build GRCh38
rs779663654 Chr.4: 39838013-39838013 on Build GRCh38
rs750921310 Chr.4: 39837933-39837933 on Build GRCh38
rs776087249 Chr.4: 39838024-39838024 on Build GRCh38
rs773720637 Chr.4: 39838065-39838065 on Build GRCh38
rs768125508 Chr.4: 39837924-39837924 on Build GRCh38
rs756082832 Chr.4: 39838091-39838091 on Build GRCh38
rs181933926 Chr.4: 39838076-39838076 on Build GRCh38
rs115575413 Chr.4: 39837965-39837965 on Build GRCh38
rs774740163 Chr.4: 39837913-39837913 on Build GRCh38
rs754081758 Chr.4: 39837939-39837939 on Build GRCh38
rs769446864 Chr.4: 39838008-39838008 on Build GRCh38
rs746872077 Chr.4: 39837956-39837956 on Build GRCh38
rs763310762 Chr.4: 39838051-39838051 on Build GRCh38
rs778970158 Chr.4: 39837952-39837952 on Build GRCh38
rs927800937 Chr.4: 39837979-39837979 on Build GRCh38
rs762058912 Chr.4: 39837922-39837922 on Build GRCh38
rs572018041 Chr.4: 39838085-39838085 on Build GRCh38

View Full Product Details