Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 155251816-155252089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TFB1M
Entrez Gene ID
51106
Gene Name
transcription factor B1, mitochondrial
Gene Aliases
CGI-75, CGI75, mtTFB, mtTFB1
UniGene
Hs.279908
Gene Chromosome Location
Chr.6: 155229990-155314497 on Build GRCh38
Transcripts
XM_011535871.1 XM_011535872.2 NM_001010927.2 NM_012454.3

Gene Symbol
TIAM2
Entrez Gene ID
26230
Gene Name
T-cell lymphoma invasion and metastasis 2
Gene Aliases
STEF, TIAM-2
UniGene
Hs.586279
Gene Chromosome Location
Chr.6: 155090289-155257723 on Build GRCh38
Transcripts
XM_011535871.1 XM_011535872.2 NM_001010927.2 NM_012454.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4609213 c.4113G>T p.K1371N Chr.6: 155251997-155251997 on Build GRCh38
4454112 c.2195A>G p.K732R Chr.6: 155251951-155251951 on Build GRCh38
5805504 c.4163T>A p.V1388D Chr.6: 155251960-155251960 on Build GRCh38
4454111 c.4154A>G p.K1385R Chr.6: 155251951-155251951 on Build GRCh38
1319135 c.4061-2A>T p.? Chr.6: 155251943-155251943 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149887106 Chr.6: 155251896-155251896 on Build GRCh38
rs762222789 Chr.6: 155252064-155252064 on Build GRCh38
rs767818256 Chr.6: 155252049-155252049 on Build GRCh38
rs377106927 Chr.6: 155251894-155251894 on Build GRCh38
rs769756084 Chr.6: 155251846-155251846 on Build GRCh38
rs779465528 Chr.6: 155251974-155251974 on Build GRCh38
rs886399856 Chr.6: 155251955-155251955 on Build GRCh38
rs3831214 Chr.6: 155251925-155251928 on Build GRCh38
rs773540711 Chr.6: 155251919-155251922 on Build GRCh38
rs751738804 Chr.6: 155251927-155251927 on Build GRCh38
rs751752451 Chr.6: 155252043-155252043 on Build GRCh38
rs372966542 Chr.6: 155251901-155251901 on Build GRCh38
rs769222249 Chr.6: 155251960-155251960 on Build GRCh38
rs770667052 Chr.6: 155251971-155251971 on Build GRCh38
rs759116572 Chr.6: 155252023-155252023 on Build GRCh38
rs746006844 Chr.6: 155251934-155251934 on Build GRCh38
rs764247399 Chr.6: 155251923-155251923 on Build GRCh38
rs1047861797 Chr.6: 155251948-155251948 on Build GRCh38
rs763823343 Chr.6: 155252042-155252042 on Build GRCh38
rs371371538 Chr.6: 155251958-155251958 on Build GRCh38
rs775975289 Chr.6: 155252001-155252001 on Build GRCh38
rs112602137 Chr.6: 155251872-155251872 on Build GRCh38
rs749338034 Chr.6: 155251919-155251919 on Build GRCh38
rs3011899 Chr.6: 155251852-155251852 on Build GRCh38
rs757396552 Chr.6: 155252047-155252047 on Build GRCh38
rs990539491 Chr.6: 155251878-155251878 on Build GRCh38
rs775424471 Chr.6: 155252031-155252031 on Build GRCh38
rs765148131 Chr.6: 155252027-155252027 on Build GRCh38
rs111273707 Chr.6: 155252063-155252063 on Build GRCh38
rs762672755 Chr.6: 155252039-155252039 on Build GRCh38
rs200239140 Chr.6: 155251967-155251967 on Build GRCh38
rs780771831 Chr.6: 155251954-155251954 on Build GRCh38
rs746664519 Chr.6: 155251968-155251968 on Build GRCh38
rs781150338 Chr.6: 155251930-155251930 on Build GRCh38
rs146614447 Chr.6: 155251959-155251959 on Build GRCh38
rs756655114 Chr.6: 155251938-155251938 on Build GRCh38
rs937772289 Chr.6: 155251856-155251856 on Build GRCh38
rs757317183 Chr.6: 155251928-155251928 on Build GRCh38
rs758250272 Chr.6: 155251903-155251903 on Build GRCh38

View Full Product Details