Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 113766138-113766383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RSBN1
Entrez Gene ID
54665
Gene Name
round spermatid basic protein 1
Gene Aliases
ROSBIN
UniGene
Hs.486285
Gene Chromosome Location
Chr.1: 113761832-113812476 on Build GRCh38
Transcripts
NM_018364.4 NR_130896.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7187334 c.2156A>G p.N719S Chr.1: 113766233-113766233 on Build GRCh38
5792096 c.2153C>G p.S718C Chr.1: 113766236-113766236 on Build GRCh38
674276 c.2058C>G p.F686L Chr.1: 113766331-113766331 on Build GRCh38
3984107 c.2036C>G p.P679R Chr.1: 113766353-113766353 on Build GRCh38
674275 c.2031C>T p.F677F Chr.1: 113766358-113766358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1027534938 Chr.1: 113766172-113766172 on Build GRCh38
rs754797156 Chr.1: 113766289-113766289 on Build GRCh38
rs755996201 Chr.1: 113766259-113766259 on Build GRCh38
rs764410365 Chr.1: 113766236-113766236 on Build GRCh38
rs541033126 Chr.1: 113766319-113766319 on Build GRCh38
rs776113007 Chr.1: 113766323-113766323 on Build GRCh38
rs749828600 Chr.1: 113766334-113766334 on Build GRCh38
rs952832309 Chr.1: 113766173-113766173 on Build GRCh38
rs746307326 Chr.1: 113766209-113766209 on Build GRCh38
rs750508575 Chr.1: 113766254-113766254 on Build GRCh38
rs747818881 Chr.1: 113766183-113766183 on Build GRCh38
rs370806258 Chr.1: 113766166-113766166 on Build GRCh38
rs139632000 Chr.1: 113766222-113766222 on Build GRCh38
rs766279820 Chr.1: 113766264-113766264 on Build GRCh38
rs778596373 Chr.1: 113766180-113766180 on Build GRCh38
rs770457454 Chr.1: 113766212-113766212 on Build GRCh38
rs772859480 Chr.1: 113766186-113766186 on Build GRCh38
rs775951282 Chr.1: 113766215-113766215 on Build GRCh38
rs771593313 Chr.1: 113766184-113766184 on Build GRCh38
rs375049710 Chr.1: 113766310-113766310 on Build GRCh38
rs572026739 Chr.1: 113766221-113766221 on Build GRCh38
rs1010489434 Chr.1: 113766261-113766262 on Build GRCh38
rs767577413 Chr.1: 113766253-113766253 on Build GRCh38
rs756280745 Chr.1: 113766248-113766248 on Build GRCh38
rs757987908 Chr.1: 113766311-113766311 on Build GRCh38
rs774629446 Chr.1: 113766242-113766242 on Build GRCh38
rs777380467 Chr.1: 113766316-113766316 on Build GRCh38
rs749101594 Chr.1: 113766163-113766163 on Build GRCh38
rs753813023 Chr.1: 113766284-113766284 on Build GRCh38
rs778628399 Chr.1: 113766307-113766307 on Build GRCh38

View Full Product Details