Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 319315-319588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPH3AL
Entrez Gene ID
9501
Gene Name
rabphilin 3A-like (without C2 domains)
Gene Aliases
NOC2
UniGene
Hs.651925
Gene Chromosome Location
Chr.17: 212389-386254 on Build GRCh38
Transcripts
NM_001190411.1 NM_001190412.1 NM_001190413.1 NM_006987.3 XM_011535364.2 XM_011535365.2 XM_011535366.2 XM_011535367.2 XM_011535368.2 XM_011535369.2 XM_017025367.1 XM_017025368.1 XM_017025369.1 XM_017025370.1 XM_017025371.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
230325 c.253C>T p.R85W Chr.17: 319518-319518 on Build GRCh38
3514702 c.241G>A p.E81K Chr.17: 319530-319530 on Build GRCh38
4722582 c.298_299insG p.E100fs*40 Chr.17: 319472-319473 on Build GRCh38
4541352 c.298G>A p.E100K Chr.17: 319473-319473 on Build GRCh38
2739890 c.254G>A p.R85Q Chr.17: 319517-319517 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763992577 Chr.17: 319542-319542 on Build GRCh38
rs1009808230 Chr.17: 319367-319367 on Build GRCh38
rs956326380 Chr.17: 319459-319459 on Build GRCh38
rs758943991 Chr.17: 319518-319518 on Build GRCh38
rs933775460 Chr.17: 319361-319361 on Build GRCh38
rs777184439 Chr.17: 319506-319506 on Build GRCh38
rs775864312 Chr.17: 319412-319412 on Build GRCh38
rs779012576 Chr.17: 319362-319362 on Build GRCh38
rs756139812 Chr.17: 319374-319374 on Build GRCh38
rs772363685 Chr.17: 319485-319485 on Build GRCh38
rs780233536 Chr.17: 319496-319496 on Build GRCh38
rs78066818 Chr.17: 319529-319529 on Build GRCh38
rs138916532 Chr.17: 319477-319477 on Build GRCh38
rs762397455 Chr.17: 319516-319516 on Build GRCh38
rs532892676 Chr.17: 319464-319464 on Build GRCh38
rs547679373 Chr.17: 319404-319404 on Build GRCh38
rs975640511 Chr.17: 319537-319537 on Build GRCh38
rs771037345 Chr.17: 319394-319394 on Build GRCh38
rs372642059 Chr.17: 319380-319380 on Build GRCh38
rs780325558 Chr.17: 319554-319554 on Build GRCh38
rs764754742 Chr.17: 319410-319417 on Build GRCh38
rs997926810 Chr.17: 319347-319347 on Build GRCh38
rs752392880 Chr.17: 319535-319535 on Build GRCh38
rs376207640 Chr.17: 319415-319415 on Build GRCh38
rs756062023 Chr.17: 319411-319411 on Build GRCh38
rs373780238 Chr.17: 319447-319447 on Build GRCh38
rs111499596 Chr.17: 319470-319470 on Build GRCh38
rs867538179 Chr.17: 319499-319499 on Build GRCh38
rs568286873 Chr.17: 319548-319548 on Build GRCh38
rs767227281 Chr.17: 319452-319452 on Build GRCh38
rs779060099 Chr.17: 319400-319400 on Build GRCh38
rs141106137 Chr.17: 319456-319456 on Build GRCh38
rs148017340 Chr.17: 319445-319445 on Build GRCh38
rs529643437 Chr.17: 319349-319349 on Build GRCh38
rs777923487 Chr.17: 319474-319474 on Build GRCh38
rs897825681 Chr.17: 319345-319345 on Build GRCh38
rs746113618 Chr.17: 319402-319402 on Build GRCh38
rs571563640 Chr.17: 319559-319559 on Build GRCh38
rs753801034 Chr.17: 319475-319475 on Build GRCh38
rs760605460 Chr.17: 319536-319536 on Build GRCh38
rs757322931 Chr.17: 319541-319542 on Build GRCh38
rs765477945 Chr.17: 319562-319563 on Build GRCh38
rs769007945 Chr.17: 319505-319505 on Build GRCh38
rs77710215 Chr.17: 319366-319366 on Build GRCh38
rs758443013 Chr.17: 319553-319553 on Build GRCh38
rs967908769 Chr.17: 319379-319379 on Build GRCh38
rs143101166 Chr.17: 319493-319493 on Build GRCh38
rs777094860 Chr.17: 319444-319444 on Build GRCh38
rs753509259 Chr.17: 319543-319543 on Build GRCh38
rs189870962 Chr.17: 319501-319501 on Build GRCh38
rs561889935 Chr.17: 319336-319336 on Build GRCh38
rs200439017 Chr.17: 319547-319547 on Build GRCh38
rs1022209459 Chr.17: 319369-319369 on Build GRCh38
rs910019978 Chr.17: 319520-319520 on Build GRCh38
rs777827218 Chr.17: 319375-319375 on Build GRCh38
rs150241223 Chr.17: 319476-319476 on Build GRCh38
rs773930646 Chr.17: 319517-319517 on Build GRCh38
rs199595016 Chr.17: 319558-319558 on Build GRCh38
rs769200302 Chr.17: 319424-319424 on Build GRCh38

View Full Product Details