Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 216859690-216859955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNP1
Entrez Gene ID
7141
Gene Name
transition protein 1
Gene Aliases
TP1
UniGene
Hs.3017
Gene Chromosome Location
Chr.2: 216859458-216860059 on Build GRCh38
Transcripts
NM_003284.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5416399 c.139G>A p.A47T Chr.2: 216859896-216859896 on Build GRCh38
1016252 c.98A>G p.Y33C Chr.2: 216859937-216859937 on Build GRCh38
1016251 c.100C>T p.R34C Chr.2: 216859935-216859935 on Build GRCh38
1016250 c.127C>T p.R43W Chr.2: 216859908-216859908 on Build GRCh38
4624566 c.139G>T p.A47S Chr.2: 216859896-216859896 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577756162 Chr.2: 216859833-216859833 on Build GRCh38
rs373054076 Chr.2: 216859748-216859748 on Build GRCh38
rs770240966 Chr.2: 216859916-216859916 on Build GRCh38
rs772553656 Chr.2: 216859923-216859923 on Build GRCh38
rs772178924 Chr.2: 216859863-216859863 on Build GRCh38
rs767789499 Chr.2: 216859889-216859890 on Build GRCh38
rs1056226612 Chr.2: 216859798-216859798 on Build GRCh38
rs756861746 Chr.2: 216859711-216859711 on Build GRCh38
rs1050474871 Chr.2: 216859740-216859740 on Build GRCh38
rs756419428 Chr.2: 216859862-216859863 on Build GRCh38
rs779115272 Chr.2: 216859915-216859915 on Build GRCh38
rs986968684 Chr.2: 216859911-216859911 on Build GRCh38
rs1022506860 Chr.2: 216859823-216859823 on Build GRCh38
rs368298412 Chr.2: 216859902-216859902 on Build GRCh38
rs780910696 Chr.2: 216859897-216859897 on Build GRCh38
rs777265915 Chr.2: 216859831-216859831 on Build GRCh38
rs200478604 Chr.2: 216859887-216859887 on Build GRCh38
rs766630762 Chr.2: 216859872-216859872 on Build GRCh38
rs576145762 Chr.2: 216859773-216859773 on Build GRCh38
rs775577081 Chr.2: 216859895-216859896 on Build GRCh38
rs768841294 Chr.2: 216859735-216859735 on Build GRCh38
rs762266940 Chr.2: 216859847-216859847 on Build GRCh38
rs561435538 Chr.2: 216859908-216859908 on Build GRCh38
rs762829245 Chr.2: 216859877-216859877 on Build GRCh38
rs749666432 Chr.2: 216859729-216859729 on Build GRCh38
rs752377768 Chr.2: 216859772-216859772 on Build GRCh38
rs769559893 Chr.2: 216859726-216859726 on Build GRCh38
rs536380970 Chr.2: 216859725-216859725 on Build GRCh38
rs531949711 Chr.2: 216859900-216859900 on Build GRCh38
rs757241176 Chr.2: 216859895-216859895 on Build GRCh38
rs778477228 Chr.2: 216859728-216859728 on Build GRCh38
rs754585675 Chr.2: 216859721-216859722 on Build GRCh38
rs184176050 Chr.2: 216859866-216859866 on Build GRCh38
rs895240743 Chr.2: 216859817-216859817 on Build GRCh38
rs774940448 Chr.2: 216859744-216859744 on Build GRCh38
rs549619953 Chr.2: 216859903-216859903 on Build GRCh38
rs760944000 Chr.2: 216859870-216859870 on Build GRCh38
rs367585957 Chr.2: 216859907-216859907 on Build GRCh38
rs780822708 Chr.2: 216859715-216859715 on Build GRCh38
rs745566654 Chr.2: 216859719-216859719 on Build GRCh38
rs62180545 Chr.2: 216859821-216859821 on Build GRCh38
rs560143746 Chr.2: 216859886-216859886 on Build GRCh38
rs753949356 Chr.2: 216859876-216859876 on Build GRCh38
rs562997102 Chr.2: 216859829-216859829 on Build GRCh38
rs554582205 Chr.2: 216859761-216859761 on Build GRCh38
rs199683943 Chr.2: 216859922-216859922 on Build GRCh38

View Full Product Details