Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 86229659-86229930 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC7A13
Entrez Gene ID
157724
Gene Name
solute carrier family 7 member 13
Gene Aliases
AGT-1, AGT1, XAT2
UniGene
Hs.149095
Gene Chromosome Location
Chr.8: 86213909-86230380 on Build GRCh38
Transcripts
NM_138817.2 XM_011516867.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5616018 c.496C>A p.L166M Chr.8: 86229782-86229782 on Build GRCh38
2789613 c.506C>T p.S169F Chr.8: 86229772-86229772 on Build GRCh38
4727763 c.372T>C p.F124F Chr.8: 86229906-86229906 on Build GRCh38
3699191 c.433G>A p.V145I Chr.8: 86229845-86229845 on Build GRCh38
3779391 c.516C>T p.S172S Chr.8: 86229762-86229762 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759113712 Chr.8: 86229774-86229774 on Build GRCh38
rs754302977 Chr.8: 86229711-86229711 on Build GRCh38
rs764378002 Chr.8: 86229710-86229710 on Build GRCh38
rs762350458 Chr.8: 86229799-86229799 on Build GRCh38
rs780882826 Chr.8: 86229740-86229740 on Build GRCh38
rs745475338 Chr.8: 86229834-86229834 on Build GRCh38
rs866577087 Chr.8: 86229707-86229707 on Build GRCh38
rs762327138 Chr.8: 86229851-86229851 on Build GRCh38
rs868695711 Chr.8: 86229905-86229905 on Build GRCh38
rs770555645 Chr.8: 86229768-86229768 on Build GRCh38
rs144944362 Chr.8: 86229826-86229826 on Build GRCh38
rs773764264 Chr.8: 86229832-86229832 on Build GRCh38
rs865879997 Chr.8: 86229791-86229791 on Build GRCh38
rs185522605 Chr.8: 86229906-86229906 on Build GRCh38
rs765911104 Chr.8: 86229800-86229800 on Build GRCh38
rs750519862 Chr.8: 86229804-86229804 on Build GRCh38
rs766574140 Chr.8: 86229817-86229817 on Build GRCh38
rs756037423 Chr.8: 86229823-86229823 on Build GRCh38
rs368361188 Chr.8: 86229822-86229822 on Build GRCh38
rs372246476 Chr.8: 86229865-86229865 on Build GRCh38
rs758496717 Chr.8: 86229812-86229812 on Build GRCh38
rs776880618 Chr.8: 86229780-86229780 on Build GRCh38
rs770359622 Chr.8: 86229828-86229828 on Build GRCh38
rs765358074 Chr.8: 86229730-86229730 on Build GRCh38
rs866841258 Chr.8: 86229900-86229900 on Build GRCh38
rs777049284 Chr.8: 86229849-86229849 on Build GRCh38
rs891536339 Chr.8: 86229763-86229763 on Build GRCh38
rs939313112 Chr.8: 86229712-86229712 on Build GRCh38
rs763589149 Chr.8: 86229862-86229862 on Build GRCh38
rs534501229 Chr.8: 86229716-86229716 on Build GRCh38
rs759777224 Chr.8: 86229896-86229896 on Build GRCh38
rs377741883 Chr.8: 86229703-86229703 on Build GRCh38
rs766520997 Chr.8: 86229881-86229881 on Build GRCh38
rs200414005 Chr.8: 86229766-86229766 on Build GRCh38
rs758689854 Chr.8: 86229731-86229731 on Build GRCh38
rs140320705 Chr.8: 86229758-86229758 on Build GRCh38
rs1051211573 Chr.8: 86229783-86229783 on Build GRCh38
rs990311495 Chr.8: 86229893-86229893 on Build GRCh38
rs747210422 Chr.8: 86229737-86229737 on Build GRCh38
rs752520540 Chr.8: 86229907-86229907 on Build GRCh38
rs761897892 Chr.8: 86229905-86229906 on Build GRCh38
rs754738072 Chr.8: 86229738-86229738 on Build GRCh38
rs762567364 Chr.8: 86229697-86229697 on Build GRCh38
rs376438148 Chr.8: 86229742-86229742 on Build GRCh38
rs755181378 Chr.8: 86229821-86229821 on Build GRCh38
rs556669007 Chr.8: 86229778-86229778 on Build GRCh38
rs142954513 Chr.8: 86229818-86229818 on Build GRCh38
rs748074011 Chr.8: 86229754-86229754 on Build GRCh38
rs867679086 Chr.8: 86229827-86229827 on Build GRCh38
rs181461170 Chr.8: 86229844-86229844 on Build GRCh38
rs370985157 Chr.8: 86229704-86229704 on Build GRCh38
rs751642802 Chr.8: 86229882-86229882 on Build GRCh38
rs768375259 Chr.8: 86229772-86229773 on Build GRCh38
rs868602680 Chr.8: 86229734-86229734 on Build GRCh38
rs116610214 Chr.8: 86229736-86229736 on Build GRCh38
rs774405787 Chr.8: 86229816-86229816 on Build GRCh38
rs773790475 Chr.8: 86229860-86229860 on Build GRCh38
rs770297736 Chr.8: 86229852-86229852 on Build GRCh38

View Full Product Details