Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 42338540-42338769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLB
Entrez Gene ID
5423
Gene Name
DNA polymerase beta
Gene Aliases
-
UniGene
Hs.654484
Gene Chromosome Location
Chr.8: 42338452-42371813 on Build GRCh38
Transcripts
NM_002690.2 XM_005273535.3 XM_005273536.3 XM_005273537.3 XM_005273538.2 XM_005273539.2 XM_005273540.4 XM_006716353.2 XM_017013583.1 XM_017013584.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3374930 c.49G>A p.D17N Chr.8: 42338673-42338673 on Build GRCh38
6231523 c.1A>G p.M1V Chr.8: 42338625-42338625 on Build GRCh38
5358720 c.31C>G p.L11V Chr.8: 42338655-42338655 on Build GRCh38
4716379 c.54G>A p.M18I Chr.8: 42338678-42338678 on Build GRCh38
1100008 c.45C>T p.I15I Chr.8: 42338669-42338669 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs780281312 Chr.8: 42338583-42338583 on Build GRCh38
rs374798020 Chr.8: 42338578-42338578 on Build GRCh38
rs771780389 Chr.8: 42338703-42338703 on Build GRCh38
rs779243575 Chr.8: 42338577-42338577 on Build GRCh38
rs2307164 Chr.8: 42338690-42338690 on Build GRCh38
rs903711999 Chr.8: 42338700-42338700 on Build GRCh38
rs763403727 Chr.8: 42338731-42338731 on Build GRCh38
rs2307160 Chr.8: 42338563-42338563 on Build GRCh38
rs545525035 Chr.8: 42338620-42338620 on Build GRCh38
rs372261806 Chr.8: 42338570-42338570 on Build GRCh38
rs766759933 Chr.8: 42338737-42338737 on Build GRCh38
rs780407939 Chr.8: 42338662-42338662 on Build GRCh38
rs778587622 Chr.8: 42338699-42338699 on Build GRCh38
rs759115794 Chr.8: 42338601-42338601 on Build GRCh38
rs745663987 Chr.8: 42338702-42338702 on Build GRCh38
rs773256437 Chr.8: 42338592-42338592 on Build GRCh38
rs767099838 Chr.8: 42338610-42338610 on Build GRCh38
rs369062208 Chr.8: 42338695-42338695 on Build GRCh38
rs200636493 Chr.8: 42338647-42338647 on Build GRCh38
rs1043536555 Chr.8: 42338688-42338688 on Build GRCh38
rs1056486121 Chr.8: 42338589-42338589 on Build GRCh38
rs746100208 Chr.8: 42338580-42338580 on Build GRCh38
rs763381943 Chr.8: 42338574-42338574 on Build GRCh38
rs747128905 Chr.8: 42338663-42338663 on Build GRCh38
rs768177415 Chr.8: 42338707-42338707 on Build GRCh38
rs775090925 Chr.8: 42338704-42338704 on Build GRCh38
rs776167021 Chr.8: 42338722-42338722 on Build GRCh38
rs762226684 Chr.8: 42338723-42338723 on Build GRCh38
rs1050145021 Chr.8: 42338747-42338747 on Build GRCh38
rs765908494 Chr.8: 42338594-42338594 on Build GRCh38
rs755089897 Chr.8: 42338669-42338669 on Build GRCh38
rs757732811 Chr.8: 42338625-42338625 on Build GRCh38
rs372427707 Chr.8: 42338603-42338603 on Build GRCh38
rs369833174 Chr.8: 42338595-42338595 on Build GRCh38
rs758602721 Chr.8: 42338582-42338582 on Build GRCh38
rs747024971 Chr.8: 42338584-42338584 on Build GRCh38
rs774160240 Chr.8: 42338597-42338597 on Build GRCh38
rs373698557 Chr.8: 42338727-42338727 on Build GRCh38
rs750729543 Chr.8: 42338656-42338656 on Build GRCh38
rs781376620 Chr.8: 42338681-42338681 on Build GRCh38
rs749219468 Chr.8: 42338685-42338685 on Build GRCh38
rs760175427 Chr.8: 42338619-42338619 on Build GRCh38
rs769906597 Chr.8: 42338585-42338585 on Build GRCh38
rs775041792 Chr.8: 42338612-42338612 on Build GRCh38
rs754319030 Chr.8: 42338622-42338622 on Build GRCh38
rs760226419 Chr.8: 42338706-42338706 on Build GRCh38
rs758560481 Chr.8: 42338661-42338661 on Build GRCh38
rs756995926 Chr.8: 42338714-42338716 on Build GRCh38

View Full Product Details