Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41866868-41867141 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SREBF2
Entrez Gene ID
6721
Gene Name
sterol regulatory element binding transcription factor 2
Gene Aliases
SREBP-2, SREBP2, bHLHd2
UniGene
Hs.443258
Gene Chromosome Location
Chr.22: 41833079-41907308 on Build GRCh38
Transcripts
NM_004599.3 NR_103834.1 XM_006724310.2 XM_011530347.1 XM_017028921.1 XM_017028922.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6210095 c.180_181insT p.G61fs*2 Chr.22: 41866922-41866923 on Build GRCh38
3554989 c.357C>T p.T119T Chr.22: 41867099-41867099 on Build GRCh38
3371926 c.236G>A p.S79N Chr.22: 41866978-41866978 on Build GRCh38
1714431 c.352C>T p.P118S Chr.22: 41867094-41867094 on Build GRCh38
3964365 c.140C>A p.S47* Chr.22: 41866882-41866882 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138407302 Chr.22: 41867085-41867085 on Build GRCh38
rs150093664 Chr.22: 41866912-41866912 on Build GRCh38
rs771095016 Chr.22: 41866926-41866946 on Build GRCh38
rs375812398 Chr.22: 41867030-41867030 on Build GRCh38
rs755835024 Chr.22: 41866986-41866986 on Build GRCh38
rs756783538 Chr.22: 41867098-41867098 on Build GRCh38
rs145442703 Chr.22: 41866934-41866934 on Build GRCh38
rs755316524 Chr.22: 41867092-41867092 on Build GRCh38
rs1051145016 Chr.22: 41866950-41866950 on Build GRCh38
rs758741776 Chr.22: 41866953-41866954 on Build GRCh38
rs765909582 Chr.22: 41866944-41866945 on Build GRCh38
rs777395605 Chr.22: 41866924-41866924 on Build GRCh38
rs758831419 Chr.22: 41866975-41866975 on Build GRCh38
rs981133162 Chr.22: 41866909-41866909 on Build GRCh38
rs763605883 Chr.22: 41867079-41867079 on Build GRCh38
rs765717734 Chr.22: 41866968-41866968 on Build GRCh38
rs747110517 Chr.22: 41866925-41866926 on Build GRCh38
rs996862586 Chr.22: 41866949-41866949 on Build GRCh38
rs139293286 Chr.22: 41866958-41866958 on Build GRCh38
rs369078163 Chr.22: 41867054-41867054 on Build GRCh38
rs760943913 Chr.22: 41866953-41866953 on Build GRCh38
rs956042805 Chr.22: 41867077-41867077 on Build GRCh38
rs775311992 Chr.22: 41867039-41867039 on Build GRCh38
rs750971461 Chr.22: 41866969-41866969 on Build GRCh38
rs550403258 Chr.22: 41866985-41866985 on Build GRCh38
rs62636525 Chr.22: 41866977-41866977 on Build GRCh38
rs767167336 Chr.22: 41866943-41866944 on Build GRCh38
rs143202499 Chr.22: 41866959-41866959 on Build GRCh38
rs757831058 Chr.22: 41866891-41866891 on Build GRCh38
rs199537318 Chr.22: 41867072-41867072 on Build GRCh38
rs917130709 Chr.22: 41867094-41867094 on Build GRCh38
rs143615881 Chr.22: 41866945-41866947 on Build GRCh38
rs538434796 Chr.22: 41866974-41866974 on Build GRCh38
rs899443374 Chr.22: 41866942-41866942 on Build GRCh38
rs779412919 Chr.22: 41866945-41866953 on Build GRCh38
rs774173456 Chr.22: 41866935-41866935 on Build GRCh38
rs571438249 Chr.22: 41866973-41866973 on Build GRCh38
rs200528058 Chr.22: 41867051-41867051 on Build GRCh38
rs775684191 Chr.22: 41866946-41866946 on Build GRCh38
rs771839575 Chr.22: 41867031-41867031 on Build GRCh38
rs879108464 Chr.22: 41867106-41867106 on Build GRCh38
rs879204876 Chr.22: 41866966-41866966 on Build GRCh38
rs779626156 Chr.22: 41866927-41866935 on Build GRCh38
rs371828133 Chr.22: 41867009-41867009 on Build GRCh38
rs753378983 Chr.22: 41866945-41866950 on Build GRCh38
rs777325974 Chr.22: 41867004-41867004 on Build GRCh38
rs2229438 Chr.22: 41867108-41867108 on Build GRCh38
rs778173987 Chr.22: 41866991-41866992 on Build GRCh38
rs776823018 Chr.22: 41866926-41866927 on Build GRCh38
rs1017402973 Chr.22: 41867038-41867038 on Build GRCh38
rs745601850 Chr.22: 41866938-41866938 on Build GRCh38
rs546801935 Chr.22: 41866943-41866943 on Build GRCh38
rs549313143 Chr.22: 41866915-41866915 on Build GRCh38
rs139624098 Chr.22: 41866917-41866917 on Build GRCh38
rs778841621 Chr.22: 41867023-41867023 on Build GRCh38
rs763739852 Chr.22: 41867095-41867095 on Build GRCh38
rs147075621 Chr.22: 41867053-41867053 on Build GRCh38
rs375061773 Chr.22: 41866980-41866980 on Build GRCh38
rs751936882 Chr.22: 41867088-41867088 on Build GRCh38
rs367980385 Chr.22: 41866982-41866982 on Build GRCh38
rs201999144 Chr.22: 41867046-41867046 on Build GRCh38
rs372419146 Chr.22: 41866892-41866892 on Build GRCh38
rs753434005 Chr.22: 41867097-41867097 on Build GRCh38
rs749153168 Chr.22: 41866929-41866929 on Build GRCh38
rs756727759 Chr.22: 41867011-41867011 on Build GRCh38
rs772191381 Chr.22: 41866920-41866940 on Build GRCh38
rs745781458 Chr.22: 41867119-41867119 on Build GRCh38
rs765301833 Chr.22: 41867070-41867070 on Build GRCh38
rs748545963 Chr.22: 41866910-41866911 on Build GRCh38
rs761600642 Chr.22: 41866935-41866936 on Build GRCh38
rs528551323 Chr.22: 41866918-41866918 on Build GRCh38
rs774079185 Chr.22: 41866929-41866930 on Build GRCh38
rs780927068 Chr.22: 41866916-41866916 on Build GRCh38
rs761888547 Chr.22: 41867064-41867064 on Build GRCh38
rs753019049 Chr.22: 41866945-41866946 on Build GRCh38
rs758849313 Chr.22: 41866914-41866914 on Build GRCh38
rs773512331 Chr.22: 41866919-41866920 on Build GRCh38

View Full Product Details