Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 9377718-9377991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THUMPD3
Entrez Gene ID
25917
Gene Name
THUMP domain containing 3
Gene Aliases
-
UniGene
Hs.443081
Gene Chromosome Location
Chr.3: 9362842-9386791 on Build GRCh38
Transcripts
NM_001114092.1 NM_015453.2 XM_005265021.4 XM_005265022.4 XM_005265023.3 XM_005265024.4 XM_006713086.3 XM_011533570.2 XM_011533571.2 XM_017006113.1 XM_017006114.1 XM_017006115.1 XM_017006116.1 XM_017006117.1 XM_017006118.1 XM_017006119.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
129721 c.967A>G p.I323V Chr.3: 9377847-9377847 on Build GRCh38
5756379 c.1008G>C p.E336D Chr.3: 9377888-9377888 on Build GRCh38
6858311 c.939-4C>G p.? Chr.3: 9377815-9377815 on Build GRCh38
1425727 c.946G>C p.D316H Chr.3: 9377826-9377826 on Build GRCh38
3427948 c.972C>T p.V324V Chr.3: 9377852-9377852 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs771714678 Chr.3: 9377808-9377808 on Build GRCh38
rs989158333 Chr.3: 9377958-9377958 on Build GRCh38
rs901985806 Chr.3: 9377839-9377839 on Build GRCh38
rs200611720 Chr.3: 9377896-9377896 on Build GRCh38
rs773754288 Chr.3: 9377803-9377803 on Build GRCh38
rs781441513 Chr.3: 9377868-9377868 on Build GRCh38
rs1016171250 Chr.3: 9377843-9377843 on Build GRCh38
rs760365262 Chr.3: 9377908-9377908 on Build GRCh38
rs760303979 Chr.3: 9377813-9377813 on Build GRCh38
rs774954171 Chr.3: 9377907-9377907 on Build GRCh38
rs750485589 Chr.3: 9377835-9377835 on Build GRCh38
rs759836045 Chr.3: 9377879-9377879 on Build GRCh38
rs765175458 Chr.3: 9377823-9377823 on Build GRCh38
rs768317771 Chr.3: 9377914-9377914 on Build GRCh38
rs1031823999 Chr.3: 9377844-9377844 on Build GRCh38
rs1055151538 Chr.3: 9377806-9377806 on Build GRCh38
rs931081112 Chr.3: 9377746-9377746 on Build GRCh38
rs753626197 Chr.3: 9377804-9377804 on Build GRCh38
rs1005540866 Chr.3: 9377833-9377833 on Build GRCh38
rs551029050 Chr.3: 9377798-9377798 on Build GRCh38
rs371834291 Chr.3: 9377918-9377918 on Build GRCh38
rs374584327 Chr.3: 9377877-9377877 on Build GRCh38
rs201378265 Chr.3: 9377867-9377867 on Build GRCh38
rs957976811 Chr.3: 9377952-9377952 on Build GRCh38
rs780277347 Chr.3: 9377837-9377837 on Build GRCh38
rs140401860 Chr.3: 9377803-9377804 on Build GRCh38
rs769210091 Chr.3: 9377785-9377785 on Build GRCh38
rs780367132 Chr.3: 9377780-9377780 on Build GRCh38
rs761614702 Chr.3: 9377919-9377919 on Build GRCh38
rs369740119 Chr.3: 9377861-9377861 on Build GRCh38
rs750478730 Chr.3: 9377770-9377770 on Build GRCh38
rs778273635 Chr.3: 9377889-9377889 on Build GRCh38
rs747384348 Chr.3: 9377782-9377782 on Build GRCh38
rs993295386 Chr.3: 9377891-9377891 on Build GRCh38
rs758718647 Chr.3: 9377772-9377772 on Build GRCh38
rs753641621 Chr.3: 9377819-9377819 on Build GRCh38
rs986306282 Chr.3: 9377920-9377920 on Build GRCh38
rs111448116 Chr.3: 9377759-9377759 on Build GRCh38
rs771624827 Chr.3: 9377902-9377902 on Build GRCh38
rs906624770 Chr.3: 9377801-9377801 on Build GRCh38
rs146391666 Chr.3: 9377921-9377921 on Build GRCh38
rs770295073 Chr.3: 9377802-9377802 on Build GRCh38
rs58292063 Chr.3: 9377822-9377822 on Build GRCh38
rs151334180 Chr.3: 9377852-9377852 on Build GRCh38
rs867812854 Chr.3: 9377939-9377939 on Build GRCh38
rs371115859 Chr.3: 9377853-9377853 on Build GRCh38
rs138446214 Chr.3: 9377883-9377883 on Build GRCh38
rs757087741 Chr.3: 9377820-9377821 on Build GRCh38
rs750454542 Chr.3: 9377926-9377926 on Build GRCh38
rs751888218 Chr.3: 9377845-9377845 on Build GRCh38
rs758486711 Chr.3: 9377836-9377836 on Build GRCh38
rs1021602317 Chr.3: 9377888-9377888 on Build GRCh38
rs945565708 Chr.3: 9377771-9377771 on Build GRCh38
rs758987627 Chr.3: 9377804-9377804 on Build GRCh38
rs781608056 Chr.3: 9377796-9377796 on Build GRCh38

View Full Product Details