Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101346436-101346709 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TIMM8A
Entrez Gene ID
1678
Gene Name
translocase of inner mitochondrial membrane 8 homolog A (yeast)
Gene Aliases
DDP, DDP1, DFN1, MTS, TIM8
UniGene
Hs.447877
Gene Chromosome Location
Chr.X: 101345656-101348969 on Build GRCh38
Transcripts
NM_004085.3

SNP Detail

SNP ID Chromosome Location
rs782484989 Chr.X: 101346574-101346574 on Build GRCh38
rs782801850 Chr.X: 101346469-101346469 on Build GRCh38
rs201897402 Chr.X: 101346542-101346542 on Build GRCh38
rs1054894 Chr.X: 101346555-101346555 on Build GRCh38
rs80356560 Chr.X: 101346595-101346595 on Build GRCh38
rs782762708 Chr.X: 101346470-101346470 on Build GRCh38
rs185711695 Chr.X: 101346474-101346474 on Build GRCh38
rs782049199 Chr.X: 101346541-101346541 on Build GRCh38
rs781878654 Chr.X: 101346587-101346587 on Build GRCh38
rs782344951 Chr.X: 101346577-101346577 on Build GRCh38
rs1054892 Chr.X: 101346549-101346549 on Build GRCh38
rs869320733 Chr.X: 101346636-101346645 on Build GRCh38
rs3027652 Chr.X: 101346514-101346514 on Build GRCh38
rs781823241 Chr.X: 101346554-101346554 on Build GRCh38
rs782690796 Chr.X: 101346642-101346642 on Build GRCh38
rs139950658 Chr.X: 101346611-101346611 on Build GRCh38

View Full Product Details