Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 1305206-1305461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNPH
Entrez Gene ID
9751
Gene Name
syntaphilin
Gene Aliases
-
UniGene
Hs.323833
Gene Chromosome Location
Chr.20: 1266291-1309327 on Build GRCh38
Transcripts
NM_001318234.1 NM_014723.3 XM_005260889.3 XM_011529402.2 XM_011529403.2 XM_011529404.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5365916 c.696C>T p.P232P Chr.20: 1305265-1305265 on Build GRCh38
4096668 c.768C>T p.D256D Chr.20: 1305337-1305337 on Build GRCh38
722655 c.853G>A p.A285T Chr.20: 1305290-1305290 on Build GRCh38
2703047 c.995C>T p.P332L Chr.20: 1305432-1305432 on Build GRCh38
5365917 c.828C>T p.P276P Chr.20: 1305265-1305265 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372413123 Chr.20: 1305271-1305271 on Build GRCh38
rs544589713 Chr.20: 1305281-1305281 on Build GRCh38
rs776133925 Chr.20: 1305231-1305231 on Build GRCh38
rs751832991 Chr.20: 1305255-1305255 on Build GRCh38
rs189950046 Chr.20: 1305355-1305355 on Build GRCh38
rs756254539 Chr.20: 1305274-1305274 on Build GRCh38
rs200706722 Chr.20: 1305361-1305361 on Build GRCh38
rs537448470 Chr.20: 1305364-1305364 on Build GRCh38
rs753820108 Chr.20: 1305329-1305329 on Build GRCh38
rs147296928 Chr.20: 1305414-1305414 on Build GRCh38
rs760887496 Chr.20: 1305325-1305325 on Build GRCh38
rs761763707 Chr.20: 1305430-1305430 on Build GRCh38
rs144751280 Chr.20: 1305330-1305330 on Build GRCh38
rs113261920 Chr.20: 1305397-1305397 on Build GRCh38
rs140499293 Chr.20: 1305256-1305256 on Build GRCh38
rs745444756 Chr.20: 1305300-1305300 on Build GRCh38
rs191018281 Chr.20: 1305249-1305249 on Build GRCh38
rs750221003 Chr.20: 1305438-1305438 on Build GRCh38
rs752657489 Chr.20: 1305278-1305278 on Build GRCh38
rs779050665 Chr.20: 1305275-1305275 on Build GRCh38
rs201060812 Chr.20: 1305367-1305367 on Build GRCh38
rs765203773 Chr.20: 1305425-1305425 on Build GRCh38
rs150545078 Chr.20: 1305385-1305385 on Build GRCh38
rs3795139 Chr.20: 1305289-1305289 on Build GRCh38
rs141041990 Chr.20: 1305317-1305317 on Build GRCh38
rs181513642 Chr.20: 1305334-1305334 on Build GRCh38
rs780818477 Chr.20: 1305298-1305298 on Build GRCh38
rs767334781 Chr.20: 1305373-1305373 on Build GRCh38
rs759710638 Chr.20: 1305424-1305424 on Build GRCh38
rs866586809 Chr.20: 1305238-1305238 on Build GRCh38
rs758979785 Chr.20: 1305232-1305232 on Build GRCh38
rs1049898539 Chr.20: 1305321-1305321 on Build GRCh38
rs775603255 Chr.20: 1305429-1305429 on Build GRCh38
rs141030371 Chr.20: 1305426-1305426 on Build GRCh38
rs776704297 Chr.20: 1305420-1305420 on Build GRCh38
rs776019533 Chr.20: 1305302-1305302 on Build GRCh38
rs779502696 Chr.20: 1305389-1305389 on Build GRCh38
rs748601042 Chr.20: 1305346-1305346 on Build GRCh38
rs746725748 Chr.20: 1305294-1305294 on Build GRCh38
rs186046719 Chr.20: 1305340-1305340 on Build GRCh38
rs529959698 Chr.20: 1305314-1305314 on Build GRCh38
rs748633621 Chr.20: 1305406-1305406 on Build GRCh38
rs143448228 Chr.20: 1305348-1305348 on Build GRCh38
rs147549046 Chr.20: 1305248-1305248 on Build GRCh38
rs754631738 Chr.20: 1305312-1305312 on Build GRCh38
rs369350508 Chr.20: 1305313-1305313 on Build GRCh38
rs376994362 Chr.20: 1305272-1305272 on Build GRCh38
rs141691964 Chr.20: 1305371-1305371 on Build GRCh38
rs200385437 Chr.20: 1305337-1305337 on Build GRCh38
rs866713734 Chr.20: 1305330-1305331 on Build GRCh38
rs769275894 Chr.20: 1305301-1305301 on Build GRCh38
rs763549663 Chr.20: 1305309-1305309 on Build GRCh38
rs778626429 Chr.20: 1305328-1305328 on Build GRCh38
rs150050410 Chr.20: 1305413-1305413 on Build GRCh38
rs1056354544 Chr.20: 1305343-1305343 on Build GRCh38
rs373433553 Chr.20: 1305333-1305333 on Build GRCh38
rs758363052 Chr.20: 1305409-1305409 on Build GRCh38
rs968214220 Chr.20: 1305263-1305263 on Build GRCh38
rs750660876 Chr.20: 1305324-1305324 on Build GRCh38
rs147974294 Chr.20: 1305363-1305363 on Build GRCh38
rs145008832 Chr.20: 1305296-1305296 on Build GRCh38
rs567905357 Chr.20: 1305386-1305386 on Build GRCh38
rs536948162 Chr.20: 1305396-1305396 on Build GRCh38
rs770353515 Chr.20: 1305247-1305247 on Build GRCh38
rs138783054 Chr.20: 1305338-1305338 on Build GRCh38
rs73579357 Chr.20: 1305339-1305339 on Build GRCh38

View Full Product Details