Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 76194974-76195201 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SV2C
Entrez Gene ID
22987
Gene Name
synaptic vesicle glycoprotein 2C
Gene Aliases
-
UniGene
Hs.646953
Gene Chromosome Location
Chr.5: 75847463-76353939 on Build GRCh38
Transcripts
NM_001297716.1 NM_014979.3 XM_011543281.2 XM_011543282.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6849022 c.704C>T p.S235F Chr.5: 76195042-76195042 on Build GRCh38
3073803 c.654A>G p.K218K Chr.5: 76194992-76194992 on Build GRCh38
6846003 c.709T>C p.F237L Chr.5: 76195047-76195047 on Build GRCh38
4731021 c.757T>A p.F253I Chr.5: 76195095-76195095 on Build GRCh38
3617906 c.719G>A p.G240D Chr.5: 76195057-76195057 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751393518 Chr.5: 76195108-76195108 on Build GRCh38
rs199971429 Chr.5: 76195022-76195022 on Build GRCh38
rs1054322733 Chr.5: 76195161-76195161 on Build GRCh38
rs965720468 Chr.5: 76195095-76195095 on Build GRCh38
rs766503936 Chr.5: 76195078-76195078 on Build GRCh38
rs756955373 Chr.5: 76194997-76194997 on Build GRCh38
rs780371830 Chr.5: 76195023-76195023 on Build GRCh38
rs780686289 Chr.5: 76195113-76195113 on Build GRCh38
rs60550320 Chr.5: 76195111-76195111 on Build GRCh38
rs756528271 Chr.5: 76195064-76195065 on Build GRCh38
rs750107812 Chr.5: 76195118-76195118 on Build GRCh38
rs373154009 Chr.5: 76195013-76195013 on Build GRCh38
rs202144317 Chr.5: 76195028-76195028 on Build GRCh38
rs762226050 Chr.5: 76195052-76195052 on Build GRCh38
rs199642813 Chr.5: 76195048-76195048 on Build GRCh38
rs762990822 Chr.5: 76195097-76195097 on Build GRCh38
rs200529235 Chr.5: 76195081-76195081 on Build GRCh38
rs757157390 Chr.5: 76195110-76195110 on Build GRCh38
rs780963041 Chr.5: 76195012-76195012 on Build GRCh38
rs1004838534 Chr.5: 76195004-76195004 on Build GRCh38
rs200453284 Chr.5: 76195067-76195067 on Build GRCh38
rs915732407 Chr.5: 76195171-76195171 on Build GRCh38
rs1028310476 Chr.5: 76195063-76195063 on Build GRCh38
rs779620246 Chr.5: 76195146-76195146 on Build GRCh38
rs755539417 Chr.5: 76195131-76195131 on Build GRCh38
rs754765131 Chr.5: 76195144-76195144 on Build GRCh38
rs893148235 Chr.5: 76195141-76195141 on Build GRCh38
rs760653251 Chr.5: 76195073-76195073 on Build GRCh38
rs773527691 Chr.5: 76195071-76195071 on Build GRCh38
rs763925349 Chr.5: 76195104-76195104 on Build GRCh38
rs935824290 Chr.5: 76195122-76195122 on Build GRCh38
rs774708883 Chr.5: 76195050-76195050 on Build GRCh38

View Full Product Details