Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 102425090-102425337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TECPR2
Entrez Gene ID
9895
Gene Name
tectonin beta-propeller repeat containing 2
Gene Aliases
KIAA0329, SPG49
UniGene
Hs.195667
Gene Chromosome Location
Chr.14: 102362912-102502481 on Build GRCh38
Transcripts
NM_001172631.2 NM_014844.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4049198 c.863A>G p.H288R Chr.14: 102425203-102425203 on Build GRCh38
3667614 c.757G>A p.A253T Chr.14: 102425097-102425097 on Build GRCh38
5499203 c.762G>A p.T254T Chr.14: 102425102-102425102 on Build GRCh38
7415706 c.793G>A p.V265I Chr.14: 102425133-102425133 on Build GRCh38
3493908 c.805G>A p.E269K Chr.14: 102425145-102425145 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201798383 Chr.14: 102425217-102425217 on Build GRCh38
rs762907209 Chr.14: 102425125-102425125 on Build GRCh38
rs745728811 Chr.14: 102425308-102425308 on Build GRCh38
rs761090703 Chr.14: 102425277-102425277 on Build GRCh38
rs776523980 Chr.14: 102425224-102425224 on Build GRCh38
rs746798798 Chr.14: 102425190-102425190 on Build GRCh38
rs770891190 Chr.14: 102425219-102425219 on Build GRCh38
rs768580023 Chr.14: 102425236-102425236 on Build GRCh38
rs917867269 Chr.14: 102425129-102425129 on Build GRCh38
rs201360354 Chr.14: 102425187-102425187 on Build GRCh38
rs761887165 Chr.14: 102425244-102425244 on Build GRCh38
rs762272406 Chr.14: 102425130-102425130 on Build GRCh38
rs754426151 Chr.14: 102425178-102425178 on Build GRCh38
rs775785898 Chr.14: 102425192-102425201 on Build GRCh38
rs764592554 Chr.14: 102425296-102425296 on Build GRCh38
rs368955042 Chr.14: 102425303-102425303 on Build GRCh38
rs756747141 Chr.14: 102425155-102425155 on Build GRCh38
rs369410138 Chr.14: 102425156-102425156 on Build GRCh38
rs758113494 Chr.14: 102425304-102425304 on Build GRCh38
rs144069125 Chr.14: 102425158-102425158 on Build GRCh38
rs760865454 Chr.14: 102425302-102425305 on Build GRCh38
rs764422441 Chr.14: 102425126-102425126 on Build GRCh38
rs754519207 Chr.14: 102425293-102425293 on Build GRCh38
rs12587242 Chr.14: 102425306-102425306 on Build GRCh38
rs758522721 Chr.14: 102425127-102425127 on Build GRCh38
rs921971593 Chr.14: 102425194-102425194 on Build GRCh38
rs371752089 Chr.14: 102425241-102425241 on Build GRCh38
rs767842978 Chr.14: 102425256-102425256 on Build GRCh38
rs199885993 Chr.14: 102425215-102425215 on Build GRCh38
rs767043358 Chr.14: 102425281-102425281 on Build GRCh38
rs61744777 Chr.14: 102425144-102425144 on Build GRCh38
rs4906197 Chr.14: 102425309-102425309 on Build GRCh38
rs932143139 Chr.14: 102425214-102425214 on Build GRCh38
rs754252362 Chr.14: 102425288-102425288 on Build GRCh38
rs553398813 Chr.14: 102425305-102425305 on Build GRCh38
rs781094413 Chr.14: 102425210-102425210 on Build GRCh38
rs374689078 Chr.14: 102425273-102425273 on Build GRCh38
rs758225483 Chr.14: 102425189-102425189 on Build GRCh38
rs775608857 Chr.14: 102425123-102425123 on Build GRCh38
rs749405948 Chr.14: 102425234-102425234 on Build GRCh38
rs936655898 Chr.14: 102425168-102425168 on Build GRCh38
rs201691545 Chr.14: 102425186-102425186 on Build GRCh38
rs375630293 Chr.14: 102425151-102425151 on Build GRCh38

View Full Product Details