Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 156767916-156768183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDGFC
Entrez Gene ID
56034
Gene Name
platelet derived growth factor C
Gene Aliases
FALLOTEIN, SCDGF
UniGene
Hs.570855
Gene Chromosome Location
Chr.4: 156761611-156971394 on Build GRCh38
Transcripts
NM_016205.2 NR_036641.1 XM_011532124.2 XM_017008455.1 XM_017008456.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4168012 c.758G>A p.R253H Chr.4: 156767936-156767936 on Build GRCh38
3129856 c.734T>C p.V245A Chr.4: 156767960-156767960 on Build GRCh38
3974785 c.769G>A p.V257M Chr.4: 156767925-156767925 on Build GRCh38
5712297 c.754C>A p.P252T Chr.4: 156767940-156767940 on Build GRCh38
6743115 c.729G>T p.E243D Chr.4: 156767965-156767965 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749851184 Chr.4: 156768014-156768014 on Build GRCh38
rs1015365643 Chr.4: 156768056-156768056 on Build GRCh38
rs753609093 Chr.4: 156768092-156768092 on Build GRCh38
rs778443868 Chr.4: 156768061-156768061 on Build GRCh38
rs760452946 Chr.4: 156767990-156767990 on Build GRCh38
rs546336037 Chr.4: 156768116-156768116 on Build GRCh38
rs777064864 Chr.4: 156767943-156767943 on Build GRCh38
rs201389930 Chr.4: 156767973-156767973 on Build GRCh38
rs770043440 Chr.4: 156767955-156767955 on Build GRCh38
rs111394094 Chr.4: 156768029-156768029 on Build GRCh38
rs773136318 Chr.4: 156767965-156767965 on Build GRCh38
rs775357932 Chr.4: 156767983-156767983 on Build GRCh38
rs780361412 Chr.4: 156768024-156768024 on Build GRCh38
rs966527659 Chr.4: 156768142-156768142 on Build GRCh38
rs371527708 Chr.4: 156767994-156767994 on Build GRCh38
rs528149454 Chr.4: 156768096-156768096 on Build GRCh38
rs534929639 Chr.4: 156768143-156768143 on Build GRCh38
rs748367828 Chr.4: 156767946-156767946 on Build GRCh38
rs149581531 Chr.4: 156768103-156768103 on Build GRCh38
rs763862874 Chr.4: 156767993-156767993 on Build GRCh38
rs1003941407 Chr.4: 156768046-156768046 on Build GRCh38
rs371877120 Chr.4: 156768034-156768034 on Build GRCh38
rs764754185 Chr.4: 156768009-156768009 on Build GRCh38
rs759551032 Chr.4: 156767967-156767967 on Build GRCh38
rs1022059233 Chr.4: 156768139-156768139 on Build GRCh38
rs572564410 Chr.4: 156768115-156768115 on Build GRCh38
rs763800226 Chr.4: 156768082-156768082 on Build GRCh38
rs149543962 Chr.4: 156767944-156767944 on Build GRCh38
rs138037357 Chr.4: 156768062-156768062 on Build GRCh38
rs200467587 Chr.4: 156768021-156768021 on Build GRCh38
rs368111342 Chr.4: 156768028-156768028 on Build GRCh38

View Full Product Details