Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 134377553-134377826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHF6
Entrez Gene ID
84295
Gene Name
PHD finger protein 6
Gene Aliases
BFLS, BORJ, CENP-31
UniGene
Hs.356501
Gene Chromosome Location
Chr.X: 134373312-134428792 on Build GRCh38
Transcripts
NM_001015877.1 NM_032335.3 NM_032458.2

SNP Detail

SNP ID Chromosome Location
rs371510284 Chr.X: 134377766-134377766 on Build GRCh38
rs758842701 Chr.X: 134377683-134377683 on Build GRCh38
rs398124428 Chr.X: 134377620-134377620 on Build GRCh38
rs132630301 Chr.X: 134377639-134377639 on Build GRCh38
rs758791658 Chr.X: 134377638-134377639 on Build GRCh38
rs132630299 Chr.X: 134377751-134377751 on Build GRCh38
rs755705850 Chr.X: 134377582-134377582 on Build GRCh38
rs748760985 Chr.X: 134377631-134377631 on Build GRCh38
rs750155732 Chr.X: 134377782-134377782 on Build GRCh38
rs755797851 Chr.X: 134377790-134377790 on Build GRCh38
rs767314368 Chr.X: 134377780-134377780 on Build GRCh38
rs367754027 Chr.X: 134377678-134377678 on Build GRCh38
rs892911660 Chr.X: 134377758-134377758 on Build GRCh38
rs140590075 Chr.X: 134377701-134377701 on Build GRCh38
rs573685409 Chr.X: 134377740-134377740 on Build GRCh38
rs1019968355 Chr.X: 134377789-134377789 on Build GRCh38
rs779548130 Chr.X: 134377626-134377626 on Build GRCh38
rs765201747 Chr.X: 134377764-134377764 on Build GRCh38
rs760978695 Chr.X: 134377739-134377739 on Build GRCh38
rs778255925 Chr.X: 134377765-134377765 on Build GRCh38
rs371190526 Chr.X: 134377651-134377651 on Build GRCh38
rs132630300 Chr.X: 134377619-134377619 on Build GRCh38
rs759708359 Chr.X: 134377749-134377749 on Build GRCh38
rs776870994 Chr.X: 134377743-134377743 on Build GRCh38
rs1057519064 Chr.X: 134377730-134377730 on Build GRCh38
rs769068524 Chr.X: 134377641-134377641 on Build GRCh38
rs376022686 Chr.X: 134377664-134377664 on Build GRCh38
rs774974810 Chr.X: 134377650-134377650 on Build GRCh38

View Full Product Details