Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45816650-45816875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBE2C
Entrez Gene ID
11065
Gene Name
ubiquitin conjugating enzyme E2 C
Gene Aliases
UBCH10, dJ447F3.2
UniGene
Hs.93002
Gene Chromosome Location
Chr.20: 45812576-45816957 on Build GRCh38
Transcripts
NM_001281741.1 NM_001281742.1 NM_007019.3 NM_181799.2 NM_181800.2 NM_181801.3 NR_104036.1 NR_104037.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1565758 c.482-1G>C p.? Chr.20: 45816708-45816708 on Build GRCh38
4738667 c.447A>G p.Q149Q Chr.20: 45816728-45816728 on Build GRCh38
5891549 c.428-4C>T p.? Chr.20: 45816705-45816705 on Build GRCh38
4738666 c.501A>G p.Q167Q Chr.20: 45816728-45816728 on Build GRCh38
1632318 c.457T>C p.S153P Chr.20: 45816738-45816738 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202149476 Chr.20: 45816732-45816732 on Build GRCh38
rs947731541 Chr.20: 45816710-45816710 on Build GRCh38
rs546303107 Chr.20: 45816688-45816688 on Build GRCh38
rs6065895 Chr.20: 45816764-45816764 on Build GRCh38
rs367766260 Chr.20: 45816824-45816824 on Build GRCh38
rs751367029 Chr.20: 45816768-45816768 on Build GRCh38
rs562364655 Chr.20: 45816812-45816812 on Build GRCh38
rs774676127 Chr.20: 45816709-45816709 on Build GRCh38
rs781461068 Chr.20: 45816769-45816769 on Build GRCh38
rs760365326 Chr.20: 45816719-45816719 on Build GRCh38
rs775305257 Chr.20: 45816687-45816688 on Build GRCh38
rs763411603 Chr.20: 45816692-45816692 on Build GRCh38
rs753350058 Chr.20: 45816726-45816726 on Build GRCh38
rs187608783 Chr.20: 45816737-45816737 on Build GRCh38
rs985255663 Chr.20: 45816760-45816760 on Build GRCh38
rs772269337 Chr.20: 45816813-45816813 on Build GRCh38
rs770260108 Chr.20: 45816685-45816685 on Build GRCh38
rs1029457888 Chr.20: 45816727-45816727 on Build GRCh38
rs748254538 Chr.20: 45816776-45816776 on Build GRCh38
rs771400267 Chr.20: 45816811-45816811 on Build GRCh38
rs115972142 Chr.20: 45816795-45816795 on Build GRCh38
rs576550569 Chr.20: 45816791-45816791 on Build GRCh38
rs766325761 Chr.20: 45816757-45816757 on Build GRCh38
rs373830041 Chr.20: 45816796-45816796 on Build GRCh38
rs376878469 Chr.20: 45816748-45816748 on Build GRCh38
rs941837194 Chr.20: 45816803-45816803 on Build GRCh38
rs911174531 Chr.20: 45816777-45816777 on Build GRCh38
rs367674756 Chr.20: 45816817-45816817 on Build GRCh38
rs955166544 Chr.20: 45816744-45816744 on Build GRCh38
rs976107634 Chr.20: 45816701-45816701 on Build GRCh38
rs933282828 Chr.20: 45816829-45816829 on Build GRCh38
rs771308119 Chr.20: 45816700-45816700 on Build GRCh38
rs748253814 Chr.20: 45816683-45816683 on Build GRCh38
rs762886220 Chr.20: 45816799-45816800 on Build GRCh38
rs758357409 Chr.20: 45816750-45816750 on Build GRCh38
rs763735948 Chr.20: 45816725-45816725 on Build GRCh38
rs761402285 Chr.20: 45816729-45816729 on Build GRCh38

View Full Product Details