Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 32931701-32931956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
QSER1
Entrez Gene ID
79832
Gene Name
glutamine and serine rich 1
Gene Aliases
-
UniGene
Hs.369368
Gene Chromosome Location
Chr.11: 32892826-32980270 on Build GRCh38
Transcripts
NM_001076786.2 XM_006718325.3 XM_017018327.1 XM_017018328.1 XM_017018329.1 XM_017018330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6132510 c.199T>A p.F67I Chr.11: 32931844-32931844 on Build GRCh38
926408 c.145C>T p.P49S Chr.11: 32931790-32931790 on Build GRCh38
5457507 c.169_170insTC p.S57fs*38 Chr.11: 32931814-32931815 on Build GRCh38
2114707 c.151A>G p.T51A Chr.11: 32931796-32931796 on Build GRCh38
3446820 c.107C>T p.S36F Chr.11: 32931752-32931752 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs907261959 Chr.11: 32931797-32931797 on Build GRCh38
rs557548317 Chr.11: 32931841-32931841 on Build GRCh38
rs767909825 Chr.11: 32931820-32931820 on Build GRCh38
rs577613656 Chr.11: 32931863-32931863 on Build GRCh38
rs983242034 Chr.11: 32931786-32931786 on Build GRCh38
rs376731888 Chr.11: 32931834-32931834 on Build GRCh38
rs755908153 Chr.11: 32931912-32931912 on Build GRCh38
rs907602281 Chr.11: 32931837-32931837 on Build GRCh38
rs751573883 Chr.11: 32931870-32931870 on Build GRCh38
rs991138733 Chr.11: 32931744-32931744 on Build GRCh38
rs754839303 Chr.11: 32931776-32931776 on Build GRCh38
rs757368488 Chr.11: 32931853-32931853 on Build GRCh38
rs781195205 Chr.11: 32931904-32931904 on Build GRCh38
rs754930684 Chr.11: 32931897-32931897 on Build GRCh38
rs746860720 Chr.11: 32931771-32931771 on Build GRCh38
rs777497808 Chr.11: 32931928-32931928 on Build GRCh38
rs764206703 Chr.11: 32931749-32931749 on Build GRCh38
rs780042245 Chr.11: 32931764-32931764 on Build GRCh38
rs770290878 Chr.11: 32931790-32931790 on Build GRCh38
rs774946336 Chr.11: 32931791-32931791 on Build GRCh38
rs375726391 Chr.11: 32931745-32931745 on Build GRCh38
rs775809153 Chr.11: 32931821-32931821 on Build GRCh38
rs764371304 Chr.11: 32931832-32931832 on Build GRCh38
rs752781916 Chr.11: 32931858-32931858 on Build GRCh38
rs747956806 Chr.11: 32931908-32931908 on Build GRCh38
rs760944139 Chr.11: 32931825-32931825 on Build GRCh38
rs149280265 Chr.11: 32931807-32931807 on Build GRCh38
rs372918831 Chr.11: 32931780-32931780 on Build GRCh38
rs758333024 Chr.11: 32931762-32931762 on Build GRCh38
rs201411044 Chr.11: 32931758-32931758 on Build GRCh38
rs781090443 Chr.11: 32931778-32931778 on Build GRCh38
rs769560066 Chr.11: 32931779-32931779 on Build GRCh38

View Full Product Details