Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 121045500-121045767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBCEL
Entrez Gene ID
219899
Gene Name
tubulin folding cofactor E like
Gene Aliases
El, LRRC35
UniGene
Hs.504136
Gene Chromosome Location
Chr.11: 121024094-121090776 on Build GRCh38
Transcripts
NM_001130047.1 NM_152715.3 XM_017017326.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3981343 c.23G>C p.S8T Chr.11: 121045713-121045713 on Build GRCh38
6797725 c.73C>T p.R25C Chr.11: 121045763-121045763 on Build GRCh38
4018649 c.22A>C p.S8R Chr.11: 121045712-121045712 on Build GRCh38
3808593 c.43G>C p.E15Q Chr.11: 121045733-121045733 on Build GRCh38
5015057 c.73C>A p.R25S Chr.11: 121045763-121045763 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs940194223 Chr.11: 121045704-121045704 on Build GRCh38
rs977209895 Chr.11: 121045540-121045540 on Build GRCh38
rs947447765 Chr.11: 121045555-121045555 on Build GRCh38
rs374983544 Chr.11: 121045644-121045644 on Build GRCh38
rs778617093 Chr.11: 121045689-121045689 on Build GRCh38
rs753641764 Chr.11: 121045674-121045674 on Build GRCh38
rs1035728777 Chr.11: 121045634-121045634 on Build GRCh38
rs188811860 Chr.11: 121045563-121045563 on Build GRCh38
rs139246650 Chr.11: 121045573-121045573 on Build GRCh38
rs925699375 Chr.11: 121045543-121045543 on Build GRCh38
rs879037922 Chr.11: 121045649-121045649 on Build GRCh38
rs548112869 Chr.11: 121045586-121045586 on Build GRCh38
rs950326848 Chr.11: 121045600-121045600 on Build GRCh38
rs756929756 Chr.11: 121045675-121045675 on Build GRCh38
rs891984292 Chr.11: 121045528-121045528 on Build GRCh38
rs757726920 Chr.11: 121045530-121045530 on Build GRCh38
rs372493054 Chr.11: 121045738-121045738 on Build GRCh38
rs769264906 Chr.11: 121045720-121045720 on Build GRCh38
rs566240623 Chr.11: 121045628-121045628 on Build GRCh38
rs747470552 Chr.11: 121045717-121045717 on Build GRCh38
rs745386122 Chr.11: 121045692-121045692 on Build GRCh38
rs1045863862 Chr.11: 121045623-121045623 on Build GRCh38
rs771642949 Chr.11: 121045700-121045700 on Build GRCh38
rs371866641 Chr.11: 121045527-121045527 on Build GRCh38
rs780739140 Chr.11: 121045701-121045701 on Build GRCh38
rs777490830 Chr.11: 121045662-121045662 on Build GRCh38
rs916142121 Chr.11: 121045591-121045591 on Build GRCh38

View Full Product Details