Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 60952928-60953187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PUS10
Entrez Gene ID
150962
Gene Name
pseudouridylate synthase 10
Gene Aliases
CCDC139, DOBI
UniGene
Hs.368348
Gene Chromosome Location
Chr.2: 60938352-61018283 on Build GRCh38
Transcripts
NM_001322123.1 NM_001322124.1 NM_001322127.1 NM_144709.3 XM_011532568.2 XM_011532570.1 XM_011532571.1 XM_011532572.1 XM_011532573.2 XM_011532574.2 XM_011532576.2 XM_011532578.1 XM_017003428.1 XM_017003429.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5397197 c.1308+1G>A p.? Chr.2: 60952996-60952996 on Build GRCh38
6760397 c.1295T>G p.L432R Chr.2: 60953010-60953010 on Build GRCh38
7313260 c.1191-6G>T p.? Chr.2: 60953120-60953120 on Build GRCh38
7138914 c.1282G>C p.D428H Chr.2: 60953023-60953023 on Build GRCh38
3734430 c.1253T>G p.I418S Chr.2: 60953052-60953052 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1007114631 Chr.2: 60953051-60953051 on Build GRCh38
rs542149861 Chr.2: 60953126-60953126 on Build GRCh38
rs765128930 Chr.2: 60953074-60953074 on Build GRCh38
rs768676551 Chr.2: 60952959-60952959 on Build GRCh38
rs573372866 Chr.2: 60953037-60953037 on Build GRCh38
rs748745709 Chr.2: 60953045-60953045 on Build GRCh38
rs758786401 Chr.2: 60953022-60953022 on Build GRCh38
rs369200821 Chr.2: 60953158-60953158 on Build GRCh38
rs753994146 Chr.2: 60953150-60953153 on Build GRCh38
rs141655259 Chr.2: 60952988-60952988 on Build GRCh38
rs767696045 Chr.2: 60952976-60952976 on Build GRCh38
rs753872427 Chr.2: 60953003-60953003 on Build GRCh38
rs138283915 Chr.2: 60953036-60953036 on Build GRCh38
rs759597175 Chr.2: 60953149-60953149 on Build GRCh38
rs776705533 Chr.2: 60953133-60953133 on Build GRCh38
rs749263420 Chr.2: 60952955-60952955 on Build GRCh38
rs764035695 Chr.2: 60953152-60953153 on Build GRCh38
rs765673888 Chr.2: 60953018-60953018 on Build GRCh38
rs1053277203 Chr.2: 60952962-60952962 on Build GRCh38
rs768731663 Chr.2: 60953054-60953054 on Build GRCh38
rs753061897 Chr.2: 60953019-60953019 on Build GRCh38
rs369738950 Chr.2: 60953063-60953063 on Build GRCh38
rs963202972 Chr.2: 60953107-60953107 on Build GRCh38
rs766519441 Chr.2: 60953130-60953130 on Build GRCh38
rs771875489 Chr.2: 60953097-60953097 on Build GRCh38
rs201238304 Chr.2: 60952971-60952971 on Build GRCh38
rs766465948 Chr.2: 60953001-60953001 on Build GRCh38
rs997950091 Chr.2: 60952969-60952969 on Build GRCh38
rs772830505 Chr.2: 60953121-60953121 on Build GRCh38
rs773448158 Chr.2: 60952981-60952981 on Build GRCh38
rs774670439 Chr.2: 60953111-60953112 on Build GRCh38
rs76770165 Chr.2: 60953153-60953153 on Build GRCh38
rs779745111 Chr.2: 60953043-60953043 on Build GRCh38
rs759805118 Chr.2: 60953151-60953152 on Build GRCh38
rs774297213 Chr.2: 60952966-60952966 on Build GRCh38
rs755479545 Chr.2: 60953008-60953008 on Build GRCh38
rs753678972 Chr.2: 60953160-60953161 on Build GRCh38
rs757049534 Chr.2: 60953161-60953161 on Build GRCh38
rs780749817 Chr.2: 60953034-60953034 on Build GRCh38

View Full Product Details