Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 40139470-40139707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC8A1
Entrez Gene ID
6546
Gene Name
solute carrier family 8 member A1
Gene Aliases
NCX1
UniGene
Hs.31961
Gene Chromosome Location
Chr.2: 40112146-40512452 on Build GRCh38
Transcripts
NM_001112800.1 NM_001112801.1 NM_001112802.1 NM_001252624.1 NM_021097.2 XM_005264514.3 XM_006712081.2 XM_006712082.3 XM_006712083.3 XM_006712084.3 XM_006712085.3 XM_011533050.1 XM_011533054.2 XM_011533055.2 XM_011533056.1 XM_011533057.2 XM_017004745.1 XM_017004746.1 XM_017004747.1 XM_017004748.1 XM_017004749.1 XM_017004750.1 XM_017004751.1 XM_017004752.1 XM_017004753.1 XM_017004754.1 XM_017004755.1 XM_017004756.1 XM_017004757.1 XM_017004758.1 XM_017004759.1 XM_017004760.1 XM_017004761.1 XM_017004762.1 XM_017004763.1 NR_038441.1

Gene Symbol
SLC8A1-AS1
Entrez Gene ID
100128590
Gene Name
SLC8A1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.744988
Gene Chromosome Location
Chr.2: 39917634-40255209 on Build GRCh38
Transcripts
NM_001112800.1 NM_001112801.1 NM_001112802.1 NM_001252624.1 NM_021097.2 XM_005264514.3 XM_006712081.2 XM_006712082.3 XM_006712083.3 XM_006712084.3 XM_006712085.3 XM_011533050.1 XM_011533054.2 XM_011533055.2 XM_011533056.1 XM_011533057.2 XM_017004745.1 XM_017004746.1 XM_017004747.1 XM_017004748.1 XM_017004749.1 XM_017004750.1 XM_017004751.1 XM_017004752.1 XM_017004753.1 XM_017004754.1 XM_017004755.1 XM_017004756.1 XM_017004757.1 XM_017004758.1 XM_017004759.1 XM_017004760.1 XM_017004761.1 XM_017004762.1 XM_017004763.1 NR_038441.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
187791 c.2323G>A p.D775N Chr.2: 40139623-40139623 on Build GRCh38
4938248 c.2322G>T p.M774I Chr.2: 40139516-40139516 on Build GRCh38
4313394 c.2269C>T p.P757S Chr.2: 40139569-40139569 on Build GRCh38
4479952 c.2265C>T p.F755F Chr.2: 40139573-40139573 on Build GRCh38
4397467 c.2162-1G>A p.? Chr.2: 40139677-40139677 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759611239 Chr.2: 40139526-40139526 on Build GRCh38
rs770499303 Chr.2: 40139624-40139624 on Build GRCh38
rs773535533 Chr.2: 40139675-40139675 on Build GRCh38
rs757993523 Chr.2: 40139597-40139597 on Build GRCh38
rs760569632 Chr.2: 40139659-40139659 on Build GRCh38
rs1035021488 Chr.2: 40139651-40139651 on Build GRCh38
rs540022475 Chr.2: 40139493-40139493 on Build GRCh38
rs775508694 Chr.2: 40139646-40139646 on Build GRCh38
rs774467752 Chr.2: 40139522-40139522 on Build GRCh38
rs371485750 Chr.2: 40139631-40139631 on Build GRCh38
rs565060338 Chr.2: 40139617-40139617 on Build GRCh38
rs779659395 Chr.2: 40139602-40139602 on Build GRCh38
rs756917419 Chr.2: 40139572-40139572 on Build GRCh38
rs879144838 Chr.2: 40139645-40139645 on Build GRCh38
rs767521100 Chr.2: 40139528-40139528 on Build GRCh38
rs763128553 Chr.2: 40139514-40139514 on Build GRCh38
rs373510583 Chr.2: 40139634-40139634 on Build GRCh38
rs760613427 Chr.2: 40139549-40139549 on Build GRCh38
rs773589237 Chr.2: 40139498-40139498 on Build GRCh38
rs748692865 Chr.2: 40139615-40139615 on Build GRCh38
rs865864562 Chr.2: 40139658-40139660 on Build GRCh38
rs376027815 Chr.2: 40139540-40139540 on Build GRCh38
rs753637785 Chr.2: 40139570-40139570 on Build GRCh38
rs113881317 Chr.2: 40139618-40139618 on Build GRCh38
rs771157907 Chr.2: 40139519-40139519 on Build GRCh38
rs781045590 Chr.2: 40139660-40139660 on Build GRCh38
rs776071537 Chr.2: 40139492-40139492 on Build GRCh38
rs763827563 Chr.2: 40139567-40139567 on Build GRCh38
rs894072210 Chr.2: 40139674-40139674 on Build GRCh38
rs1021237955 Chr.2: 40139576-40139576 on Build GRCh38
rs771128762 Chr.2: 40139623-40139623 on Build GRCh38
rs199921708 Chr.2: 40139523-40139523 on Build GRCh38
rs139842575 Chr.2: 40139663-40139663 on Build GRCh38
rs137900136 Chr.2: 40139582-40139582 on Build GRCh38
rs770080096 Chr.2: 40139495-40139495 on Build GRCh38

View Full Product Details