Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 32933993-32934266 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
QSER1
Entrez Gene ID
79832
Gene Name
glutamine and serine rich 1
Gene Aliases
-
UniGene
Hs.369368
Gene Chromosome Location
Chr.11: 32892826-32980270 on Build GRCh38
Transcripts
NM_001076786.2 XM_006718325.3 XM_017018327.1 XM_017018328.1 XM_017018329.1 XM_017018330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
926418 c.2503T>G p.F835V Chr.11: 32934148-32934148 on Build GRCh38
6355073 c.2464G>T p.D822Y Chr.11: 32934109-32934109 on Build GRCh38
4481400 c.2505C>T p.F835F Chr.11: 32934150-32934150 on Build GRCh38
40850 c.2349G>A p.Q783Q Chr.11: 32933994-32933994 on Build GRCh38
4998385 c.2422C>A p.H808N Chr.11: 32934067-32934067 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778837560 Chr.11: 32934138-32934138 on Build GRCh38
rs754052538 Chr.11: 32934141-32934141 on Build GRCh38
rs771246588 Chr.11: 32934226-32934226 on Build GRCh38
rs759643917 Chr.11: 32934093-32934093 on Build GRCh38
rs767999167 Chr.11: 32934027-32934027 on Build GRCh38
rs766603318 Chr.11: 32934071-32934071 on Build GRCh38
rs764139300 Chr.11: 32934189-32934189 on Build GRCh38
rs751696976 Chr.11: 32934202-32934202 on Build GRCh38
rs987565535 Chr.11: 32934087-32934087 on Build GRCh38
rs750245972 Chr.11: 32934140-32934140 on Build GRCh38
rs920606722 Chr.11: 32934213-32934213 on Build GRCh38
rs779871760 Chr.11: 32934163-32934163 on Build GRCh38
rs781523963 Chr.11: 32934032-32934032 on Build GRCh38
rs866275654 Chr.11: 32934103-32934103 on Build GRCh38
rs374250160 Chr.11: 32934037-32934037 on Build GRCh38
rs763226460 Chr.11: 32934069-32934069 on Build GRCh38
rs746528148 Chr.11: 32934026-32934026 on Build GRCh38
rs773550937 Chr.11: 32934057-32934057 on Build GRCh38
rs751733643 Chr.11: 32934076-32934076 on Build GRCh38
rs376287754 Chr.11: 32934107-32934107 on Build GRCh38
rs748746295 Chr.11: 32934194-32934194 on Build GRCh38
rs777222714 Chr.11: 32934031-32934031 on Build GRCh38
rs773653114 Chr.11: 32934201-32934201 on Build GRCh38
rs374738343 Chr.11: 32934235-32934235 on Build GRCh38
rs763332122 Chr.11: 32934219-32934219 on Build GRCh38
rs770196397 Chr.11: 32934196-32934196 on Build GRCh38
rs746615946 Chr.11: 32934168-32934168 on Build GRCh38
rs757131827 Chr.11: 32934111-32934111 on Build GRCh38
rs770242182 Chr.11: 32934042-32934042 on Build GRCh38
rs753866520 Chr.11: 32934108-32934108 on Build GRCh38
rs200879298 Chr.11: 32934170-32934170 on Build GRCh38

View Full Product Details