Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 170996586-170996847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLK1
Entrez Gene ID
9874
Gene Name
tousled like kinase 1
Gene Aliases
PKU-beta
UniGene
Hs.744917
Gene Chromosome Location
Chr.2: 170990823-171231314 on Build GRCh38
Transcripts
NM_001136554.1 NM_001136555.1 NM_012290.4 XM_011512238.2 XM_017005421.1 XM_017005422.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7447361 c.2084T>C p.V695A Chr.2: 170996693-170996693 on Build GRCh38
6292933 c.2069T>A p.L690* Chr.2: 170996708-170996708 on Build GRCh38
7447362 c.1940T>C p.V647A Chr.2: 170996693-170996693 on Build GRCh38
6292934 c.1925T>A p.L642* Chr.2: 170996708-170996708 on Build GRCh38
1178626 c.1939G>C p.V647L Chr.2: 170996694-170996694 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755326125 Chr.2: 170996771-170996775 on Build GRCh38
rs775832565 Chr.2: 170996634-170996634 on Build GRCh38
rs1045314 Chr.2: 170996719-170996719 on Build GRCh38
rs200299178 Chr.2: 170996635-170996635 on Build GRCh38
rs781461483 Chr.2: 170996778-170996782 on Build GRCh38
rs372222009 Chr.2: 170996609-170996609 on Build GRCh38
rs764357460 Chr.2: 170996637-170996637 on Build GRCh38
rs1042841965 Chr.2: 170996640-170996640 on Build GRCh38
rs747391375 Chr.2: 170996763-170996764 on Build GRCh38
rs374595379 Chr.2: 170996631-170996631 on Build GRCh38
rs774588067 Chr.2: 170996624-170996624 on Build GRCh38
rs564340817 Chr.2: 170996816-170996816 on Build GRCh38
rs558479097 Chr.2: 170996628-170996628 on Build GRCh38
rs751437890 Chr.2: 170996792-170996792 on Build GRCh38
rs748338332 Chr.2: 170996789-170996790 on Build GRCh38
rs1053319088 Chr.2: 170996814-170996815 on Build GRCh38
rs765979797 Chr.2: 170996787-170996787 on Build GRCh38
rs750147824 Chr.2: 170996764-170996764 on Build GRCh38
rs757727208 Chr.2: 170996686-170996686 on Build GRCh38
rs777065076 Chr.2: 170996647-170996647 on Build GRCh38
rs780545999 Chr.2: 170996764-170996764 on Build GRCh38
rs764930912 Chr.2: 170996749-170996749 on Build GRCh38
rs762296537 Chr.2: 170996683-170996683 on Build GRCh38

View Full Product Details