Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 18730118-18730391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPEF1
Entrez Gene ID
5475
Gene Name
protein phosphatase with EF-hand domain 1
Gene Aliases
PP7, PPEF, PPP7C, PPP7CA
UniGene
Hs.211589
Gene Chromosome Location
Chr.X: 18675909-18827921 on Build GRCh38
Transcripts
NM_006240.2 NM_152224.1 NM_152226.1 XM_005274553.2 XM_017029612.1

SNP Detail

SNP ID Chromosome Location
rs775348591 Chr.X: 18730278-18730278 on Build GRCh38
rs771835124 Chr.X: 18730276-18730276 on Build GRCh38
rs1013765444 Chr.X: 18730269-18730269 on Build GRCh38
rs749778503 Chr.X: 18730231-18730231 on Build GRCh38
rs371822471 Chr.X: 18730352-18730352 on Build GRCh38
rs777810268 Chr.X: 18730217-18730217 on Build GRCh38
rs368328736 Chr.X: 18730340-18730340 on Build GRCh38
rs781445029 Chr.X: 18730183-18730183 on Build GRCh38
rs778281528 Chr.X: 18730228-18730228 on Build GRCh38
rs771284448 Chr.X: 18730239-18730239 on Build GRCh38
rs749409593 Chr.X: 18730335-18730335 on Build GRCh38
rs756256257 Chr.X: 18730194-18730194 on Build GRCh38
rs748215627 Chr.X: 18730193-18730193 on Build GRCh38
rs774546210 Chr.X: 18730246-18730246 on Build GRCh38
rs746282633 Chr.X: 18730252-18730252 on Build GRCh38
rs374360016 Chr.X: 18730279-18730279 on Build GRCh38
rs765261559 Chr.X: 18730347-18730347 on Build GRCh38
rs763533733 Chr.X: 18730323-18730323 on Build GRCh38
rs768064918 Chr.X: 18730312-18730312 on Build GRCh38

View Full Product Details