Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102133180-102133419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPRC1
Entrez Gene ID
23082
Gene Name
peroxisome proliferator-activated receptor gamma, coactivator-related 1
Gene Aliases
PRC
UniGene
Hs.533551
Gene Chromosome Location
Chr.10: 102132994-102150333 on Build GRCh38
Transcripts
NM_001288727.1 NM_001288728.1 NM_015062.4 XM_005269656.3 XM_006717731.3 XM_011539549.2 XM_011539551.2 XM_011539552.2 XM_017015982.1 XM_017015983.1 XM_017015984.1 XM_017015985.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4639646 c.141C>A p.S47R Chr.10: 102133209-102133209 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs974810559 Chr.10: 102133292-102133292 on Build GRCh38
rs981506015 Chr.10: 102133392-102133392 on Build GRCh38
rs1011725394 Chr.10: 102133327-102133327 on Build GRCh38
rs577687380 Chr.10: 102133308-102133308 on Build GRCh38
rs766661307 Chr.10: 102133212-102133212 on Build GRCh38
rs765593980 Chr.10: 102133246-102133246 on Build GRCh38
rs927767588 Chr.10: 102133388-102133388 on Build GRCh38
rs753005423 Chr.10: 102133247-102133247 on Build GRCh38
rs776691743 Chr.10: 102133234-102133234 on Build GRCh38
rs1036093074 Chr.10: 102133393-102133393 on Build GRCh38
rs1042199136 Chr.10: 102133363-102133363 on Build GRCh38
rs999260925 Chr.10: 102133311-102133311 on Build GRCh38
rs866613288 Chr.10: 102133213-102133213 on Build GRCh38
rs966262175 Chr.10: 102133288-102133288 on Build GRCh38
rs554932312 Chr.10: 102133197-102133197 on Build GRCh38
rs534331919 Chr.10: 102133261-102133261 on Build GRCh38
rs957353679 Chr.10: 102133317-102133317 on Build GRCh38
rs755557993 Chr.10: 102133270-102133270 on Build GRCh38
rs35674448 Chr.10: 102133366-102133366 on Build GRCh38
rs537099374 Chr.10: 102133243-102133243 on Build GRCh38
rs1031865335 Chr.10: 102133316-102133316 on Build GRCh38
rs546356937 Chr.10: 102133329-102133329 on Build GRCh38
rs774690301 Chr.10: 102133232-102133232 on Build GRCh38
rs939222495 Chr.10: 102133394-102133394 on Build GRCh38
rs552999885 Chr.10: 102133289-102133289 on Build GRCh38
rs879448469 Chr.10: 102133320-102133320 on Build GRCh38
rs573347845 Chr.10: 102133259-102133259 on Build GRCh38
rs970677035 Chr.10: 102133337-102133337 on Build GRCh38
rs933003147 Chr.10: 102133304-102133304 on Build GRCh38
rs951042111 Chr.10: 102133218-102133218 on Build GRCh38
rs902296468 Chr.10: 102133310-102133310 on Build GRCh38
rs749991163 Chr.10: 102133264-102133264 on Build GRCh38
rs773362045 Chr.10: 102133209-102133209 on Build GRCh38
rs760628604 Chr.10: 102133210-102133210 on Build GRCh38
rs945299878 Chr.10: 102133343-102133343 on Build GRCh38
rs759964857 Chr.10: 102133240-102133240 on Build GRCh38
rs1027823359 Chr.10: 102133211-102133211 on Build GRCh38
rs762058101 Chr.10: 102133380-102133380 on Build GRCh38

View Full Product Details