Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 67886926-67887185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD51B
Entrez Gene ID
5890
Gene Name
RAD51 paralog B
Gene Aliases
R51H2, RAD51L1, REC2
UniGene
Hs.172587
Gene Chromosome Location
Chr.14: 67819779-68683118 on Build GRCh38
Transcripts
NM_001321809.1 NM_001321810.1 NM_001321812.1 NM_001321814.1 NM_001321815.1 NM_001321817.1 NM_001321818.1 NM_001321819.1 NM_001321821.1 NM_002877.5 NM_133509.3 NM_133510.3 XM_011537050.2 XM_011537051.2 XM_017021545.1 XM_017021546.1 XM_017021547.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6761933 c.665C>T p.A222V Chr.14: 67887113-67887113 on Build GRCh38
7109687 c.673C>G p.Q225E Chr.14: 67887121-67887121 on Build GRCh38
7109684 c.673C>G p.Q225E Chr.14: 67887121-67887121 on Build GRCh38
6923094 c.728A>G p.K243R Chr.14: 67887176-67887176 on Build GRCh38
5228867 c.619G>C p.V207L Chr.14: 67887067-67887067 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1044316737 Chr.14: 67887002-67887002 on Build GRCh38
rs757271750 Chr.14: 67886996-67886997 on Build GRCh38
rs17104744 Chr.14: 67887162-67887162 on Build GRCh38
rs1000141790 Chr.14: 67887085-67887085 on Build GRCh38
rs758195315 Chr.14: 67886974-67886974 on Build GRCh38
rs905808360 Chr.14: 67887032-67887032 on Build GRCh38
rs377316917 Chr.14: 67887087-67887087 on Build GRCh38
rs112272290 Chr.14: 67886980-67886980 on Build GRCh38
rs775586905 Chr.14: 67887008-67887008 on Build GRCh38
rs745701150 Chr.14: 67887078-67887079 on Build GRCh38
rs770905132 Chr.14: 67886995-67886995 on Build GRCh38
rs1008737274 Chr.14: 67887012-67887012 on Build GRCh38
rs891521987 Chr.14: 67887135-67887135 on Build GRCh38
rs763308176 Chr.14: 67887017-67887017 on Build GRCh38
rs371376587 Chr.14: 67887027-67887027 on Build GRCh38
rs372664423 Chr.14: 67887080-67887080 on Build GRCh38
rs996313162 Chr.14: 67886963-67886963 on Build GRCh38
rs202221268 Chr.14: 67886988-67886988 on Build GRCh38
rs751395885 Chr.14: 67887156-67887156 on Build GRCh38
rs752481677 Chr.14: 67886970-67886970 on Build GRCh38
rs374887191 Chr.14: 67886996-67886996 on Build GRCh38
rs762382499 Chr.14: 67887047-67887047 on Build GRCh38
rs575909907 Chr.14: 67887110-67887110 on Build GRCh38
rs773288970 Chr.14: 67886971-67886975 on Build GRCh38
rs775150306 Chr.14: 67887024-67887024 on Build GRCh38
rs28908168 Chr.14: 67887067-67887067 on Build GRCh38
rs149154702 Chr.14: 67887147-67887147 on Build GRCh38
rs774910383 Chr.14: 67887040-67887040 on Build GRCh38
rs745964896 Chr.14: 67886998-67886998 on Build GRCh38
rs764878768 Chr.14: 67887082-67887082 on Build GRCh38
rs778850393 Chr.14: 67887007-67887009 on Build GRCh38
rs770007179 Chr.14: 67887006-67887006 on Build GRCh38
rs1033414164 Chr.14: 67887142-67887142 on Build GRCh38
rs369286040 Chr.14: 67887103-67887103 on Build GRCh38

View Full Product Details