Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 68188511-68188762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP3R1
Entrez Gene ID
5534
Gene Name
protein phosphatase 3 regulatory subunit B, alpha
Gene Aliases
CALNB1, CNB, CNB1
UniGene
Hs.280604
Gene Chromosome Location
Chr.2: 68178857-68252519 on Build GRCh38
Transcripts
NM_000945.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1409039 c.187G>A p.D63N Chr.2: 68188547-68188547 on Build GRCh38
7291795 c.186C>T p.F62F Chr.2: 68188548-68188548 on Build GRCh38
7385321 c.211G>C p.D71H Chr.2: 68188523-68188523 on Build GRCh38
1565509 c.71G>A p.G24E Chr.2: 68188663-68188663 on Build GRCh38
5670482 c.203G>T p.G68V Chr.2: 68188531-68188531 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879151809 Chr.2: 68188641-68188641 on Build GRCh38
rs777625092 Chr.2: 68188714-68188714 on Build GRCh38
rs140640493 Chr.2: 68188683-68188683 on Build GRCh38
rs755961823 Chr.2: 68188572-68188572 on Build GRCh38
rs772618517 Chr.2: 68188712-68188713 on Build GRCh38
rs552718073 Chr.2: 68188725-68188725 on Build GRCh38
rs114269546 Chr.2: 68188726-68188726 on Build GRCh38
rs546646019 Chr.2: 68188548-68188548 on Build GRCh38
rs757840602 Chr.2: 68188713-68188713 on Build GRCh38
rs687 Chr.2: 68188635-68188635 on Build GRCh38
rs909975831 Chr.2: 68188611-68188611 on Build GRCh38
rs747628124 Chr.2: 68188710-68188710 on Build GRCh38
rs778299520 Chr.2: 68188692-68188692 on Build GRCh38
rs866820627 Chr.2: 68188625-68188625 on Build GRCh38
rs687 Chr.2: 68188635-68188635 on Build GRCh38
rs200459875 Chr.2: 68188684-68188684 on Build GRCh38
rs4482466 Chr.2: 68188588-68188588 on Build GRCh38
rs777091669 Chr.2: 68188735-68188735 on Build GRCh38
rs763101811 Chr.2: 68188539-68188539 on Build GRCh38
rs951794466 Chr.2: 68188565-68188565 on Build GRCh38
rs751778712 Chr.2: 68188552-68188552 on Build GRCh38
rs945300597 Chr.2: 68188731-68188731 on Build GRCh38

View Full Product Details