Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15949858-15950091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR10H4
Entrez Gene ID
126541
Gene Name
olfactory receptor family 10 subfamily H member 4
Gene Aliases
OR19-28
UniGene
Hs.553579
Gene Chromosome Location
Chr.19: 15949008-15949958 on Build GRCh38
Transcripts
NM_001004465.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3529967 c.891G>A p.K297K Chr.19: 15949898-15949898 on Build GRCh38
3529968 c.892G>A p.E298K Chr.19: 15949899-15949899 on Build GRCh38
4562442 c.926G>A p.R309K Chr.19: 15949933-15949933 on Build GRCh38
3529965 c.854C>T p.P285L Chr.19: 15949861-15949861 on Build GRCh38
3891798 c.884G>C p.R295T Chr.19: 15949891-15949891 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367689468 Chr.19: 15949947-15949947 on Build GRCh38
rs757436336 Chr.19: 15949920-15949920 on Build GRCh38
rs755208785 Chr.19: 15949976-15949976 on Build GRCh38
rs774474509 Chr.19: 15949885-15949885 on Build GRCh38
rs762844145 Chr.19: 15949933-15949934 on Build GRCh38
rs371896781 Chr.19: 15949993-15949993 on Build GRCh38
rs1014276080 Chr.19: 15949914-15949914 on Build GRCh38
rs762551549 Chr.19: 15950002-15950002 on Build GRCh38
rs755901871 Chr.19: 15949961-15949961 on Build GRCh38
rs965186625 Chr.19: 15950054-15950054 on Build GRCh38
rs775169206 Chr.19: 15949960-15949960 on Build GRCh38
rs763572447 Chr.19: 15949920-15949920 on Build GRCh38
rs764883241 Chr.19: 15949912-15949912 on Build GRCh38
rs921156778 Chr.19: 15950047-15950047 on Build GRCh38
rs1000496488 Chr.19: 15949991-15949991 on Build GRCh38
rs1019482794 Chr.19: 15950021-15950021 on Build GRCh38
rs779986601 Chr.19: 15949962-15949962 on Build GRCh38
rs374132788 Chr.19: 15949944-15949944 on Build GRCh38
rs749529008 Chr.19: 15949972-15949972 on Build GRCh38
rs768313575 Chr.19: 15950004-15950004 on Build GRCh38
rs938252380 Chr.19: 15949911-15949911 on Build GRCh38
rs748223049 Chr.19: 15949990-15949990 on Build GRCh38
rs954022721 Chr.19: 15950016-15950016 on Build GRCh38
rs778902623 Chr.19: 15949981-15949981 on Build GRCh38
rs747379105 Chr.19: 15949996-15949996 on Build GRCh38
rs1594685 Chr.19: 15950017-15950017 on Build GRCh38
rs772539660 Chr.19: 15949892-15949892 on Build GRCh38
rs777042962 Chr.19: 15950000-15950000 on Build GRCh38
rs745573178 Chr.19: 15949924-15949924 on Build GRCh38
rs774938948 Chr.19: 15949915-15949915 on Build GRCh38
rs760982887 Chr.19: 15949899-15949899 on Build GRCh38
rs375793305 Chr.19: 15950006-15950006 on Build GRCh38
rs748655190 Chr.19: 15949886-15949886 on Build GRCh38
rs773601878 Chr.19: 15949898-15949898 on Build GRCh38

View Full Product Details